Search
Protein extracts isolated from cells or tissues are reduced, alkylated and digested overnight. Samples are labeled with the TMT Reagents and then mixed before sample fractionation and clean up. Labeled samples are analyzed by high resolution Orbitrap LC-MS/MS before data analysis to identify peptides and quantify reporter ion relative abundance.