PTEN Antibody (512400) in IP-MS
IP-MS enrichment of PTEN (LFQ intensity): PTEN was enriched 169-fold from MCF7 lysate compared to background proteins, using the optimized IP-MS workflow with Pierce MS-Compatible Magnetic IP Kit protein A/G (Part No. 90409) and PTEN antibody (Part No. 51-2400). See more information on IP-MS verification of antibody selectivity.
IP-MS enrichment of PTEN (LFQ intensity): PTEN was enriched 169-fold from MCF7 lysate compared to background proteins, using the optimized IP-MS workflow with Pierce MS-Compatible Magnetic IP Kit protein A/G (Part No. 90409) and PTEN antibody (Part No. 51-2400). See more information on IP-MS verification of antibody selectivity.
Related Products
Related Images