Search
POROS Analytical Chromatography Columns, available in 10- and 20-micron particle size, enable rapid separation for biomolecules for both analytical and lab-scale preparative applications.
POROS Protein A Analytical Columns are used to monitor monoclonal antibody titer and yield from cell culture supernatant. POROS CaptureSelect Analytical Columns expand the advantages of high-speed, high-performance quantitation to antibody fragments, biosimilars, and fusion proteins.
CaptureSelect conjugated ligands can be used to develop a range of analytical assays, including ELISA, Western blot, and label-free analysis. Applications include quantitation and characterization of antibody-derived therapeutics, plasma proteins, biosimilars, and even viruses and vaccines.
Learn more about POROS A, G, AL, EP, OH, and POROS CaptureSelect columns and review the list of products available for affinity, activated affinity, and metal chelate affinity chromatography.
Review products available for purification strategies using ion exchange, reversed phase chromatography, and hydrophobic interactions.
We offer an extensive list of biotinylated ligands for a wide range of applications and analytical assays.
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing:
Research use or further manufacturing. Not for diagnostic use or direct administration in humans or animals.