Search
Enabling excellent recovery for a wide range of bacteria, including anaerobes, our bacterial collection and transport solutions were designed to safely protect the integrity of your bacterial cultures.
| Catalog number | Product name |
| TS0001A | Thermo Scientific™ Amies Agar Gel Transport Swab |
Cultivate aerobic and anaerobic organisms, as well as streptococci, pneumococci, meningococci and other fastidious organisms with highly nutritious prepared media tubes.
| Catalog number | Product name |
| TV5090E | Thermo Scientific™ Brain Heart Infusion (BHI) Broth |
| TV5008D | |
| TV5001D | Thermo Scientific™ Thioglycollate Medium USP Prepared Media Tubes |
Ready-to-use, disposable inoculation loops provide enhanced recovery and reproducibility via unique gel preservation system, including convenient, ATCC®-derived strains specifically for anaerobe testing.
| Catalog number | Product name |
| R4601250 | Thermo Scientific™ Culti-Loops™ Bacteroides fragilis ATCC® 25285™ |
| R4601600 | Thermo Scientific™ Culti-Loops™ Clostridium perfringens ATCC® 13124™ |
Easily recover anaerobic bacteria including Bacteroides, Prevotella and Porphyromonas spp. with highly selective and nutritious prepared culture media plates.
Use Thermo Scientific biplate media to save incubator space and cost of atmosphere generating materials.
| Catalog number | Product name |
| Thermo Scientific™ Schaedler Anaerobe Agar Plates and Biplates: | |
| PB5034A | Schaedler Anaerobe Agar with Sheep Blood, Haemin and Vitamin K1 |
| PO5020A | Oxoid™ Schaedler Anaerobe KV Selective Agar with Lysed Horse Blood |
| PB5204E | Schaedler Anaerobe Agar / Schaedler Anaerobe KV Selective Agar biplate |
| Thermo Scientific™ A.R.I.A Medium: | |
| PB1243A | |
| PB1244A | |
Identify over 90 medically important anaerobes in just four hours using the Thermo Scientific™ RapID™ ANA II System, an identification system based on enzyme technology.
| Catalog number | Product name |
| R8311002 |
Meet new EUCAST guidelines for AST of selected rapidly growing anaerobic bacteria, with new Fastidious Anaerobe Agar with Horse Blood, combined with a variety of manual AST disks. Or, achieve minimum inhibitory concentration (MIC) results with our AST plate for testing anaerobes and B. fragilis group.
| Catalog number | Product name |
| PO10062 | NEW: Thermo Scientific™ Liofilchem Fastidious Anaerobe Agar with Horse Blood (FAA - HB) |
| ANAERO3 | Thermo Scientific™ Sensititre™ Anaerobe ANAERO3 MIC Plate |
| Thermo Scientific™ Oxoid™ AST Discs: | |
| CT0774B | |
| CT0064B | |
| CT0067B | |
| CT0152B | |
| CT0188B | |
Flexible atmosphere generation systems (AGS) in a variety of formats to support your choice of environment, including anaerobic, microaerophilic and CO2. Our end-to-end AGS range includes CE-marked jars with the convenience of one sachet and indicator to streamline your workflow while reducing waste.
| Catalog number | Product name | |
| AG0025A | Thermo Scientific™ Oxoid™ AnaeroJar™ 2.5 L Jar | |
| Thermo Scientific™ Oxoid™ AnaeroBox™: | ||
| AB0025A | ||
| AB0035A | ||
| Thermo Scientific™ Oxoid™ AnaeroGen™: | ||
| AN0025A | ||
| AN0035A | ||
| AN0010C | ||
| AN0010W | ||
Less incubator space | |
Less hands-on time | |
Less ancillary products | |
Less waste |
The European Committee on Antimicrobial Susceptibility Testing (EUCAST) has recently introduced new guidelines for performing AST of selected rapidly growing anaerobes via disc diffusion methodology on Fastidious Anaerobe Agar (FAA). Liofilchem Fastidious Anaerobe Agar with Horse Blood (FAA-HB) is a ready-to-use agar plate for the cultivation and AST of anaerobic bacteria. This medium is recommended by EUCAST for disc diffusion of selected rapidly growing anaerobic bacteria, including Bacteroides spp., Prevotella spp., Fusobacterium necrophorum, Clostridium perfringens and Cutibacterium acnes.2
Request your no-charge evaluation sample below**
Speaker: Prof. Dr. Sören Gatermann, ESCMID Fellow, Clinical Microbiologist, Chairman of the German NAC, Member of the EUCAST Steering Committee, Ruhr-Universität Bochum, Germany
This webinar addresses changes to the EUCAST breakpoint tables, valid from January 2022 (v12.0) with a focus on:
1. https://www.who.int/news-room/fact-sheets/detail/antibiotic-resistance#:~:text=Antibiotic%20resistance%20is%20accelerated%20by,poor%20infection%20prevention%20and%20control.
2. https://www.eucast.org/fileadmin/src/media/PDFs/EUCAST_files/Anaerobic_bacteria/2022_anaerobes/EUCAST_disk_diffusion_methodology_for_rapidly_growing_anaerobic_bacteria_with_QC_v_1.0_2022.pdf
Thermo Fisher Scientific is accredited by A2LA to 6559.01: ISO 17034:2016 Reference Material Producers ![]()
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Remel Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures. The identity, purity, and authenticity of the Licensed Products are exclusively the responsibility of Remel Inc., and not ATCC.
* For research use only. Not for use in diagnostic procedures.
**Terms and conditions apply.