Search
Early risk stratification in patients with (suspected) infection or sepsis is an important component of clinical evaluation and may support appropriate level-of-care considerations. However, assessing early risk and the potential for progression to a more severe disease state can be challenging, especially in patients with atypical symptoms, such as children, geriatric patients, or high-risk patients with co-morbidities as cardiac or oncological conditions.
Different clinical scores have been developed to assess disease severity and risk of adverse outcome – typically mortality, including qSOFA, NEWS, NEWS2 CURB-65 in the ED and APACHE, SOFA, SAPS in the ICU. However, the ED risk scores often lack specificity, leading to unnecessary hospitalization and increased hospital workload. ICU scores require laboratory testing and thus may not promptly capture individual organ system dysfunction1, and incorporation into daily routine is hampered by their relative complexity. Furthermore, ICU scores measure the status of all the different organs but not the risk for organ failure progression.
An increase in MR-proADM concentration has been associated with a (pathophysiological) disturbance in the microcirculation and elevation of endothelial permeability independent of infection etiology and may recognize (pending) organ dysfunction early2-3, before the appearance of clinical signs and symptoms, thereby identifying the risk for disease progression and potentially supporting timely clinical management decisions.
MR-proADM is a diagnostic parameter which support additional prognostic information and may offer improved risk stratification compared to certain standard risk scores in patients with conditions such as lower respiratory tract infections4-7, urinary tract infections and kidney disease8-10, as well as in patients suffering from sepsis, severe sepsis and septic shock.3,11,12-14 Furthermore, MR-proADM has been shown in clinical studies to be a very good predictor of outcome in patients with acute15 and chronic heart failure16-17, as well as in patients after myocardial infarction.15,18
MR-proADM may be particularly informative in patients with low to moderate SOFA scores, serving as a predictor of outcomes2, identifying the risk of disease progression early, potentially supporting timely clinical management decisions.
MR-proADM is a diagnostic parameter which can support additional prognostic information and may offer improved risk stratification, compared to certain standard risk scores in patients with conditions such as lower respiratory tract infections4-7, urinary tract infections and kidney disease8-10, as well as in patients suffering from sepsis, severe sepsis and septic shock.3,11,12-14 Furthermore, it has been shown in clinical studies that MR-proADM is a predictor of outcome in patients with acute15 and chronic heart failure16-17, as well as in patients after myocardial infarction.15,18
MR-proADM may be particularly informative in patients with low to moderate SOFA scores, serving as a strong predictor of outcomes2, identifying patients at increased risk of disease progression, potentially supporting timely clinical management decisions.
MR-proADM results can aid, in conjunction with clinical evaluation and other laboratory findings, in making decisions on level of care3,9,19-20, monitoring treatment response and making prognostic assessments for patients3,21-23 independent of the etiology of infection.24 Serial measurements can be used to monitor and track progression of the disease.3,19,25
MR-proADM concentration is low in a healthy population, with a median concentration of 0.54 nmol/L and 97.5th percentile at 0.82 nmol/L*. In patients suspected of having an infection, an elevated MR-proADM concentration, in conjunction with clinical evaluation and other laboratory findings, can assist clinicians in making decisions regarding hospital discharge (≤0.87 nmol/L), hospital admission (>0.87 nmol/L) or the potential need for a higher level of care (>1.50 nmol/L).
MR-proADM allows for additional risk assessment in patients with sepsis or septic shock and may complement standard risk assessment methods (e.g. SOFA). Serial measurements and detection of increasing values of MR-proADM can be used to monitor disease progression and treatment response.3,21-23 At the same time, a decrease in MR-proADM concentration aids in the consideration for out-patient management.26
For patients with cardiovascular disease, MR-proADM concentrations above 1.5 nmol/L can aid, along with clinical evaluation and other laboratory findings, such as NT-proBNP, troponin, and other routine biomarkers of cardiovascular disease, to assess the risk of early progression to a more severe disease state and to determine the necessity of hospital admission and level of care.27-28
*values were obtained using B·R·A·H·M·S MR-proADM II KRYPTOR assay
An observational study with 1000+ patients across 9 EDs in Europe identified and validated the 0.87 nmol/L MR-proADM cut-off in patients presenting to the ED with signs of infection. Values equal to or below 0.87 nmol/L can identify patients who may be eligible for outpatient treatment and were associated with a potential reduction of hospital admission by 15% in this population.20
Remeasure after 24 hours if clinical evaluation and MR-proADM value do not agree.
In a subsequent interventional trial in 5 EDs in Spain with 200 patients, the investigators reported that implementation of the algorithm led to a 50% increase in patients discharged (42% in SoC vs. 63% in the interventional group).26
MR-proADM values higher than 0.87 nmol/L have demonstrated a strong predictive capability in determining the necessity of hospitalization for patients with (suspected) infection. Moreover, patients with concentrations exceeding 1.5 nmol/L are associated with an elevated 28-day mortality rate (LR+ 3.47). In light of these results, a higher level of care should be considered.20
Furthermore, when used in addition to NEWS, qSOFA, and CRB-65, MR-proADM can identify patients who are at risk for disease progression, i.e. a complicated infection, which are missed by scores. An observational study reported that 89% of patients with a low clinical score, i.e. NEWS below 4, underwent an uncomplicated infection and 11% progressed to a more severe disease state of which 23% died within 28 days. Addition of MR-proADM to the risk assessment was reported to have identified these patients at hospital admission in that study population.20
The assessment of disease severity and progression became a critical focus during the COVID-19 pandemic given the scarcity of hospital resources. Numerous observational studies conducted in both ED and ICU have highlighted the potential role of MR-proADM in assessing the risk of early progression to a more severe disease state and in supporting decisions regarding the appropriate level of care.29-31 Of the biomarkers independently associated with outcome, MR-proADM had the highest hazard ratio for disease progression (6.8) and 28-day mortality (10.5), compared to D-dimer, IL-6 and Ferritin.32 When applying the low disease severity cut-off at hospital admission, the negative predictive value of MR-proADM was more than 97% for predicting in-hospital mortality.32
These studies demonstrate the reported utility of MR-proADM in viral infections, a finding that has been previously described in cases of community-acquired pneumonia.24,33
The use of MR-proADM in the emergency department can help with the assessment of further disease progression and thereby increase outpatient treatment decisions.
Disease severity assesment
MR-proADM–Guided Severity Stratification
Time-Sensitive Treatment Decisions
Beyond Traditional Scoring Systems
Sepsis has a reported hospital mortality in the ICU of 26% according to the Sepsis-3 guidelines.34 Prompt assessment of infectious load and disease severity in the early stages of sepsis is crucial for improving outcomes. Furthermore, rapid treatment with antibiotics is central, as a delay progressively increases mortality.35 Assessment of organ failure, integral to the Sepsis-3 definition, provides a measure for disease severity. Clinical scores such as SOFA, APACHE II, and SAPS II have been developed for this purpose36; however, their incorporation into daily routine is often hindered by their relative complexity. Although lactate measurement for poor perfusion is readily available, it is often too unspecific and late to be effective.
MR-proADM utility in assessing ICU patients with sepsis and septic shock was shown by Elke et al. in 2018.3 MR-proADM had the strongest association with 28-day mortality across the patient population and could facilitate a more accurate classification of low and high disease severity. Especially in patients with low to moderate SOFA scores, MR-proADM is predicative of outcomes2, identifying the risk for disease progression early and potentially supporting timely clinical management decisions. . Failure to decrease the concentration of MR-proADM in the bloodstream despite appropriate antimicrobial therapy may indicate a risk of treatment failure and may require alternative diagnosis and possible therapeutic interventions, whereas decreasing concentrations may support decisions to decrease the level of care.3
Healthcare professionals face numerous challenges in the current healthcare system, including overcrowded EDs, staff shortages, and limited hospital bed availability.37-39 These challenges necessitate an efficient decision-making process to effectively manage patient care and mitigate the risk of adverse outcomes.
One reason for ED overcrowding is the influx of patients with minor complaints that could be treated as outpatients, further straining ED resources.40-41
Notably, infections of the respiratory tract, urinary tract, intra-abdominal region, as well as skin and soft tissue infections are among the leading causes of ED admissions.42-45 For these patients, it is crucial to assess the severity of their condition and the likelihood of disease progression. This assessment is essential in determining the appropriate therapy measures to prevent the disease from advancing to more severe stages. If these infections are poorly managed, they can potentially lead to life-threatening conditions, with sepsis being the most common.20
Given these circumstances, there is a clear public need for improving the diagnostic and prognostic approach to infections in the ED. This improvement would aid supporting discharge decision making and enable the early detection of potentially at-risk patients.
In 2023, a health economic analysis was conducted using primary data from a single hospital in Spain. The study incorporated data from 473 patients to develop a model simulating 10,000 patients. These results demonstrated that MR-proADM-aided triage resulted in an admission of 41%, compared to 63.5% with standard care, which was associated with an approximate 30% cost reduction.46
The combined use of PCT and MR-proADM was part of the LIFE-POC study including more than 1,400 patients presenting with suspected organ dysfunction based on a qSOFA score greater than or equal to 1 to three large tertiary care EDs in Germany. Among those patients, 29% developed sepsis. While PCT outperformed other markers for the prediction of sepsis within 96 hours, MR-proADM showed prognostic value for in-hospital mortality. The combined use of PCT and MR-proADM demonstrated the highest predictive performance for septic shock within 96 hours in that study.48
Using PCT to aid in diagnosis and MR-proADM for prognosis has also been described previously for sepsis and septic shock.49-50
The Third International Consensus Definitions for Sepsis and Septic Shock (Sepsis-3) specified Sepsis as a life-threatening organ dysfunction caused by a dysregulated host response to infection.1,47
While B·R·A·H·M·S PCT measurements can identify and confirm a clinically suspected bacterial infection thereby guiding the initiation and duration of antibiotic treatment, B·R·A·H·M·S MR-proADM measurements help provide early identification of disease severity and progression by monitoring the development of organ dysfunction. Consequently, these measurements can support informed decisions regarding early organ support, appropriate level of care, and in-patient and out-patient treatment considerations in conjunction with clinical evaluation.
Ready to enhance your infection management strategy?
Thermo Scientific B·R·A·H·M·S KRYPTOR Analyzers are fully automated random-access benchtop immunoanalyzers that use Nobel prize winning TRACE technology to help deliver fast and reliable test results for essential indications.
Designed to support high-quality laboratory performance and patient management.
B·R·A·H·M·S biomarker solutions help support the early diagnosis and treatment of life-threatening diseases.
Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
© 2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified. B·R·A·H·M·S MR-proADM is a registered trademark of B·R·A·H·M·S GmbH.
Nobel Prize is a registered trademark of the Nobel Foundation. Other product names in this document are used for identification purposes; they may be trademarks and/or registered trademarks of their respective companies.
TRACE and KRYPTOR are trademarks of Cisbio Bioassays, licensed for use by B·R·A·H·M·S GmbH, a part of Thermo Fisher Scientific.
View patents and patent marking.