Vanquish Access HPLC System
Designed for your essential analyses, the Thermo Scientific Vanquish Access HPLC System helps prevent out-of-spec conditions, sample rework, unplanned repairs, and extra audit scrutiny.
The intelligently configured, cost-effective and fit-for-purpose Vanquish Access HPLC system delivers stable operation and trouble-free performance you can depend on.
Highlights
Robust performance
Maximize uptime with software notifications of real-time
health checks alongside built-in diagnostic and maintenance procedures.
Trouble-free method transfer
Achieve the same separations without needing to replumb or requalify with the built-in Vanquish Access HPLC System flow path tuning.
Repeatable separations
Highest flow and gradient precision and reproducibility using Thermo Scientific SmartFlow pump technology.
Delivering ease of use
Automation, compliance and networking capabilities are delivered through a tight connection to Thermo Scientific Chromeleon Chromatography Data System (CDS).
Reliable leak-free connections
Easy, tool-free column installation using Thermo Scientific Viper Fingertight Fittings.
Videos
See why you will love the Vanquish Access HPLC System.
System specifications
Category
Vanquish Access HPLC System
Download specification sheet
System pressure, maximum 500 bar
Flow range 0–10 mL/min
Pump options (default gradient delay volume contribution) Quaternary (679 µL)
Biocompatible No
pH range 1–13, chloride concentration up to 0.1 mol/L
System design Integrated, with slide-in modules
Touchscreen user interface display compatible No
Solvent monitoring system compatible No
Customizable gradient delay volume to simplify method transfer Yes, with adjustable autosampler metering device (idle volume) and method transfer kit
Injection volume range
Standard: 0.01–100 µL
Optional: 0.01–1000 µL
Sample cooling Yes, 4-40°C
Sample capacity (standard)
Based on standard 12 mm OD vials (≤ 1.5 mL)
4 racks of 54 vials (≤ 1.5 mL) each, other rack formats available
+ capacity of 12 × 22.5 mm OD vials (≤ 10 mL) in the carousel
Column compartment thermostatting Yes, 5–85°C Still air
Column compartment pre-heater Passive
Column compartment post-column cooler No
Detector Variable Wavelength Detector
Contact us to get a quote or discuss an HPLC/UHPLC solution
Use as General Laboratory Equipment only. Not for use in diagnostic procedures.