sho_evos-lightcubes
Invitrogen EVOS Light Cubes integrate LEDs and high-performance filters in a novel illumination system that delivers precise control, minimal maintenance, and exceptional reliability.
These interchangeable LED cubes provide:
  • Shorter light paths for more efficient fluorescent detection
  • Longer bulb lifetime (>50,000 hours) for lower maintenance costs
  • Adjustable light intensity to minimize photobleaching
  • Continuous light intensity for more consistent results
View the EVOS dye compatibility table
Download the EVOS dye compatibility table PDF
These LED cubes are interchangeable, have plug-and-play capability and can be easily transferred between the Invitrogen Countess II FL, EVOS FL, and Invitrogen EVOS FL Auto 2 Cell Imaging System.
Countess II FL Automated Cell Counter
Fast and accurate cell counting with fluorescence
EVOS FL Cell Imaging System
Simple fluorescence imaging with increased flexibility
Invitrogen EVOS FL Auto 2 Cell Imaging System
Time-lapse imaging at high resolution
Light Cube* Ex (nm) Em (nm) Key Compatible Dyes   
Cat. No.
DAPI 357/44 447/60 DAPI, Hoechst, BFP  AMEP4650
TagBFP 390/18 447/60 TagBFP AMEP4668
CFP 445/45 510/42 ECFP, lucifer yellow AMEP4653
GFP 470/22 510/42 GFP, Invitrogen Alexa Fluor 488, FITC, Applied Biosystems SYBR Green AMEP4651
YFP 500/24 524/27 EYFP, acridine orange (+ DNA) AMEP4654
RFP 531/40 593/40 RFP, Invitrogen Alexa Fluor 546, Alexa Fluor 555, Cy3, DsRed, Rhodamine Red AMEP4652
Invitrogen Texas Red 585/29 624/40   Invitrogen Texas Red, Alexa Fluor 568, Alexa Fluor 594, MitoTracker Red, mCherry, Cy3.5 AMEP4655
Invitrogen Cy5 628/40 692/40 Invitrogen Cy5, Alexa Fluor 647, Alexa Fluor 660, DRAQ5 AMEP4656
Invitrogen Cy5.5 655/46  794/16  Cy5.5 AMEP4673
Invitrogen Cy7 710/40 775/46 Cy7, IRDye® 800CW AMEP4667
CFP-YFP 445/45 542/27 CFP/YFP (for FRET applications) AMEP4669
AO 442/46 488LP Acridine orange (+RNA, simultaneous red/green with FL color) AMEP4670
AOred 442/46 628/32 Acridine orange + RNA, CTC formazan, Invitrogen Fura Red (high Ca2+) AMEP4671
Invitrogen Qdot 525–800 445/45 525-800 All Invitrogen Qdot nanocrystals AMEP4666
Qdot 525 445/45   525/15 Qdot 525 AMEP4657
Qdot 545 445/45  545/22 Qdot 545 AMEP4658
Qdot 565 445/45   565/16 Qdot 565 AMEP4659
Qdot 585  445/45 586/20 Qdot 585 AMEP4660
Qdot 605  445/45 605/15 Qdot 605 AMEP4661
Qdot 625 445/45   625/15 Qdot 625 AMEP4662
Qdot 655 445/45   655/15 Qdot 655 AMEP4663
Qdot 705 445/45   705/40 Qdot 705 AMEP4664
Qdot 800 445/45   785/62 Qdot 800 AMEP4665
*Click the Light Cube link for a complete list of compatible dyes/reagents.
The Power of LED Illumination
All of the EVOS fluorescence cell imaging systems utilize LED light sources. That means you get high-intensity output over a short light path for the most efficient fluorophore excitation.
  • Shorter light path provides better detection of fluorescent signals
  • Continuous illumination gives consistent results
  • >50,000-hour bulb lifetime lowers your laboratory costs
  • Adjustable light intensity reduces photobleaching
Revolutionary light path
By placing the LED light cube as close as possible to the objective turret, the number of optical elements in the light path is minimized. High-intensity illumination over a short light path increases the efficiency of fluorophore excitation, providing better detection of weak fluorescent signals.
Continuous light intensity
Mercury arc lamps can decrease in intensity by 50% in the first 100 hours of operation—plus, images acquired in different sessions cannot be quantitatively compared using mercury illumination without complicated calibrations. Because EVOS systems have continuous light cube intensity, users can rely on consistent illumination and can compare results quantitatively from images acquired on different days.
Stability Comparison
Less expensive to own and maintain
The LED bulbs on the EVOS systems are rated for >50,000 hours (~17 years), compared to 300 hours for a typical mercury bulb (and 1,500 hours for a metal halide bulb). That means significant savings in the overall upkeep of your instrument.
What light sources really cost
evos light source compare
EVOS hard-coated filter sets for higher transmission efficiency
Hard-coated filter sets are more expensive, but they have sharper edges and significantly higher transmission efficiencies that typically result in >25% more light transmission than traditional soft-coated filters. With the EVOS system’s hard-coated filter sets, your light cubes cost less over time. Plus, you will have brighter fluorescence, higher transmission efficiencies, the ability to detect faint fluorescence signals, and better signal-to-noise ratios.
    TRANSMISSION EFFICIENCY COMPARISON
Superior transmission efficiencies are observed using hard-coated filters on the EVOS instruments, compared to soft-coated filters. Excitation filter (purple), emission filter (red), dichroic mirror (green); Alexa Fluor 488 excitation (blue), Alexa Fluor 488 emission (pink).