Now offering products including extracellular matrices, scaffolds, and proteins for in vivo–like morphology and physiologically-relevant systems
AlgiMatrix™ 3D Culture System
AlgiMatrix™ 3D Culture System is the first user-friendly, animal-free bioscaffold available for the development of higher fidelity cell culture models that are more predictive of disease states and drug responses. AlgiMatrix™ 3D Culture System allows for in vivo–like morphology and better intercellular interactions.
CELLstart™ Cell Attachment Expansion Substrate
CELLstart™ is the first xeno-free substrate for attachment and expansion of human embryonic, mesenchymal, and neural stem cells that is fully-defined, has better lot-to-lot consistency, and is produced under cGMP.
Geltrex™ Basement Membranes
Geltrex™ matrix is a soluble form of reduced growth factor (RGF) basement membrane extract (BME) purified from continuous sheets of specialized extracellular matrix that form an interface between Engelbreth-Holm-Swarm (EHS) tumor cells.
Laminin is the major glycoprotein component of basement membranes, and has a molecular weight of 850,000 daltons. This preparation of mouse Laminin was isolated from the Engelbreth-Holm-Swarm (EHS) sarcoma and is supplied at a concentration of 0.5-2.0 mg⁄ml
Collagen I, Rat Tail and Collagen I, Bovine for cell culture is a high quality form of collagen widely used in 3D applications and as coating solution. Collagen I is a heterotrimeric extracellular matrix protein found in the skin and other connective tissues.
Collagen Coated Plates
High-quality, consistent coating of rat tail collagen I for culturing primary cells on plates, available in 6-well, 24-well, and 96-well formats. The plates are ready to use right from the package
Attachment Factor
A sterile solution (1x) containing gelatin as an attachment factor. When used to coat culture surfaces, AF enhances the growth of microvascular endothelial cells.