The power of custom gene synthesis is in the ability to design your DNA according to your needs and without the constraints of traditional cloning. Equally important to most researchers, however, is obtaining high yields of mRNA and ultimately protein from synthetic genes. We developed GeneArt GeneOptimizer software to help maximize expression of synthetic genes in all available expression systems. Sequence optimization is optional with every GeneArt project.
How is gene optimization different from codon optimization?
Codon optimization takes advantage of the degeneracy of the genetic code. Because of degeneracy, one protein can be encoded by many alternative nucleic acid sequences. Codon preference (codon usage bias) differs in each organism, and it can create challenges for expressing recombinant proteins in heterologous expression systems, resulting in low and unreliable expression. This may also be true for autologous expression, since wild type sequences are not necessarily optimized for expression yield but also for degradation, regulation, and other properties.
However, codon optimization is not the only relevant factor for efficient protein expression.
Our GeneOptimizer algorithm  enables true multiparametric gene optimization, dealing with a large number of sequence-related parameters involved in different aspects of gene expression, such as transcription, splicing, translation, and mRNA degradation. It considers all relevant optimization parameters in a single operation and offers a DNA sequence configured with your specifications, optimized for maximum performance in your system (Table 1).
Rationally weighing the combination of the optimization parameters (e.g., codon adaptation, mRNA de novo synthesis and stability, transcription and translation efficiency) is important to achieve the most efficient expression of a given protein.
Schematic representation of protein expression in the cell
Figure 1. Schematic representation of protein expression.
Table 1. Parameters that influence protein expression.
Transcriptional levelmRNA levelTranslational level
  • GC content
  • Consensus splice sites
  • Cryptic splice sites
  • SD sequences
  • TATA boxes
  • Termination signals
  • Artificial recombination sites
  • RNA instability motifs
  • Ribosomal entry sites
  • Repetitive sequences
  • Codon usage
  • Premature poly(A) sites
  • Ribosomal entry sites
  • Secondary structures
One further advantage of synthetic sequences is that you won’t be dependent on available DNA templates, and you can design your sequence exactly according to your requirements. Add or remove restriction sites, start/stop codons, tags, and further motifs as needed.
Sequence optimization using the GeneOptimizer software is included as an optional step with all GeneArt Gene Synthesis and DNA fragments services. To take advantage of this service, select your expression host when setting up a request using our online customer portal. The online software will then guide you through the project setup process, including sequence optimization. The resulting optimized performance, described below, is just one added value of GeneArt DNA. 
How does the GeneOptimizer process work?
Gene optimization with GeneOptimizer technology is easily performed within a few minutes using our online customer portal . Design your synthetic gene by uploading your sequence, selecting your expression system, and specifying your cloning vector and your sequence details (including open reading frames, untranslated regions, and cloning sites). Once you submit your request, the GeneOptimizer software generates the DNA sequence that best suits your research requirements, based on consideration of all parameters that are relevant for the given host organism plus your individual sequence requirements.
Use of GeneOptimizer technology results in higher protein expression
We have conducted several internal studies, and many customers have reported independently that GeneArt codon and sequence optimization results in higher protein expression without losing protein function.
In a first-of-its-kind study [1], five important protein classes were selected for optimization: protein kinases, transcription factors, ribosomal proteins, cytokines, and membrane proteins. Then 50 human genes were chosen from the NCBI database to represent the five protein classes. The selected genes were individually optimized using the GeneOptimizer algorithm [2]. For comparison, the corresponding wild type genes were subcloned using native sequences available from the NCBI database. Each gene was then expressed in triplicate in HEK293T cells. Following optimization, the 50 genes all showed reliable expression and 86% exhibited elevated expression (example in Figure 2). Further analysis showed no detrimental effect on protein solubility, and functionality was unaltered, as demonstrated for JNK1, JNK3, and CDC2 (data not shown).
Using the GeneOptimizer algorithm, in this study:
  • 86% of optimized genes showed significantly increased protein expression
  • Protein yields increased up to 15-fold with optimized genes
  • 100% of optimized genes were expressed, versus 88% of wild type genes
Western blot analysis showing protein expression in cell culture supernatants for secreted proteins or cell lysates for other proteins
Figure 2. Comparative expression analysis of wild type vs. optimized genes representing different protein classes. (A) Cell culture supernatants (for secreted proteins) or cell lysates (all other proteins) were analyzed by western blot using an anti-His antibody. One example of each protein class is shown. A 60 kDa protein used to standardize protein amount is visible, including in the empty vector negative controls. Left of each image: molecular mass values in kDa. Right of each image: identifiers for specific protein bands. (B) Relative expression levels were derived for wild type or optimized constructs (mean of three independent transfections). The fold increase in expression for the optimized construct is indicated for each protein. There was no detectable expression for IL-2 using the wild type construct. (Figure adapted from Fath et al., 2011 [1]).
Figure 3 illustrates another example of observed increases in both mRNA and protein yields of the HIV gag protein after sequence optimization using GeneOptimizer software.
Expression analysis of HIV-1 Pr55gag in transiently transfected H1299 cells using ELISA, western blot, and northern blot
Figure 3. Expression of HIV-1 Pr55gag in transiently transfected H1299 cells was analyzed by ELISA, western, and northern blot analysis. Similar results were obtained after transfection of COS7, CHO and HeLa cells. Work was performed at the University Hospital Regensburg in R. Wagner’s group [3].
To demonstrate the value of the GeneOptimizer software, we compared protein expression of sequences optimized by different vendors. A set of 10 antibody sequences was optimized with the GeneArt GeneOptimizer algorithm or one of three competitor algorithms. All constructs were transfected and expressed for (A) 7 days using the Gibco Expi293 Expression System or (B) 10 days using the Gibco ExpiCHO Expression System. Supernatant titers were measured in duplicate by biolayer interferometry (BLI) (Figure 4).
A two-panel figure showing box-and-whisker plots that compare antibody supernatant titers (produced using codon-optimized constructs supplied by Thermo Fisher Scientific and three other suppliers)
Figure 4. The GeneArt GeneOptimizer algorithm gives rise to higher antibody supernatant titers than competitor codon optimization algorithms. Lower specification limit (LSL—red dotted line) is the minimum titer needed for multiple downstream assays.
Resources for gene optimization
  1. Multiparameter RNA and codon optimization: a standardized tool to assess and enhance autologous mammalian gene expression. Fath S, Bauer AP, Liss M et al. (2011). PLoS One 6(3):e17596. doi: 10.1371/journal.pone.0017596.
  2. The GeneOptimizer Algorithm: using a sliding window approach to cope with the vast sequence space in multiparameter DNA sequence optimization. Raab D, Graf M, Notka F et al. (2010). Syst Synth Biol 4:215–225. doi: 10.1007/s11693-010-9062-3.
  3. Codon-optimized genes that enable increased heterologous expression in mammalian cells and elicit efficient immune responses in mice after vaccination of naked DNA. Graf M, Deml L, Wagner R (2004). Methods Mol Med 94:197–210. doi: 10.1385/1-59259-679-7:197 PMID: 14959831.
Request more information about GeneArt optimization and our gene synthesis service online or send us an email to geneartsupport@thermofisher.com.
Videos
Resources
Watch how-to videos and explainers on GeneArt Instant Designer.
Contact us
Request more information
North America and Asia: 1-800-955-6288
Ask for "GeneArt" when prompted
Europe: +49 (0)941 942 76-100
Order support:
Geneartsupport@thermofisher.com
Stay up to date on gene synthesis and protein synthesis service offerings.
Read white papers on de novo gene synthesis technologies.
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments