KingFisher instruments come in three different formats to meet various throughput levels and automation needs. If you need help selecting an instrument please contact your sales representative, or request a demo to see how KingFisher will work in your lab.
  KingFisher Duo Prime KingFisher Flex KingFisher Presto
Size Compact benchtop instrument Benchtop instrument Integrate with liquid handler
Max sample number/run 6/12/24 per run/load 24 or 96 96
Volume range 30-5000 µl 20-5000 µl 50-5000 µl
Plate formats
  • 24 Deep well
  • 96 Deep Well
  • Elution strip
  • 96 KF plate
  • 24 Deep well
  • 96 Deep Well
  • PCR*
  • 96 KF plate
  • 24 Deep well
  • 96 Deep Well
Heating Yes Yes Yes
Cooling Yes No No
Bar code reader With BindIt Software;
Stand alone
With BindIt Software Part of the system
UV decontamination Yes No No
Number of user selectable magnetic heads 2 4 2
Works with BindIt Software Yes Yes Yes
Stand alone use Yes Yes No, must be integrated with liquid handler. Protocol test run capability through BindIt
Request a demo
Learn More
Request a demo
Learn More
Request a demo
Learn More
*Recommended plate types only