Search
Use Thermo Scientific™ Orion™ pH Electrode Filling Solution with pH electrodes and reference electrodes. Packaged as five 2 oz. (5 x 60mL) bottles.
Orion Solutions, Standards and Buffers Overview
Ensure the proper performance and function from your electrode by using our specially formulated fill solutions
| Solution | Description | Use With Electrodes |
|---|---|---|
| 810007 | ROSS electrode fill solution, 3 M KCl, 5 x 60 mL | ROSS Ultra, ROSS and Micro ROSS Electrodes 800300, 800500U, 8102BN, 8102BNUWP, 8103BN, 8103BNUWP, 8104BN, 8104BNUWP, 8115BN, 8115BNUWP, 8135BN, 8135BNUWP, 815600, 8156BNUWP, 8157BNU, 8157BNUMD, 8157UWMMD, 8163BNWP, 8165BNWP, 8172BNWP, 8175BNWP, 8220BNWP, 8302BNUMD |
| 900011 | Silver Chloride pH electrode fill solution, 4 M KCl with Ag/AgCl, 5 x 60 mL | Standard, Specialty, Single Junction Refillable Green, Micro and ORP Electrodes 9102BNWP, 9103BNWP, 9104BNWP, 9156BNWP, 9157BN, 9157BNMD, 9162BNWP, 9165BNWP, 9172BNWP, 9180BNMD, 9678BNWP, 9778BNWP, 9780SC, 9810BN, 9826BN, 9863BN, GS9156BNWP |
| 900004 | Silver Chloride pH electrode (low level) fill solution, 2 M KCl with Ag/AgCl, 5 x 60 mL | Standard, Specialty, Single Junction Refillable Green, Micro and ORP Electrodes (low ionic strength samples) 9102BNWP, 9103BNWP, 9104BNWP, 9156BNWP, 9157BN, 9157BNMD, 9162BNWP, 9165BNWP, 9172BNWP, 9180BN, 9180BNMD, 9678BNWP, 9778BNWP, 9780SC, 9810BN, 9826BN, 9863BN, GS9156BNWP |
| 910008-WA | Double junction pH electrode fill solution, 3 M KCl, 5 x 60 mL | Double Junction, Double Junction Refillable Green and KNIpHE Electrodes 9102DJWP, 9110DJWP, 9120APWP, GD9156BNWP |
| 510011 | No Cal pH electrode fill solution, 5 x 60 mL | No Cal Electrodes 5107BNMD, 5107NC |
| 610001 | pHuture pH electrode fill solution, 5 x 60 mL | pHuture Electrodes |
| 900001 | Single junction reference electrode fill solution, equitransferent solution with Ag/AgCl, 5 x 60 mL | Single Junction Reference Electrode 900100 |
| 900002 | Double junction reference electrode inner fill solution, equitransferent solution with Ag/AgCl, 5 x 60 mL | Double Junction Reference Electrode 900200 |
| 900003 | Double junction reference electrode outer fill solution, 10 % KNO3, 5 x 60 mL | Double Junction Reference Electrode 900200 |
The ideal choice for dedicated pH testing in water, process, research, and QA/QC samples. The Thermo Scientific portfolio of pH Meters offer a range from simple & economical models to powerful & sophisticated pH meters, to help you analyze your samples with confidence.
Take your pH and ORP measurements to the field without sacrificing accuracy and reliability. Our portable meters are rugged and waterproof, and provide the dependable performance that your demanding applications require.
An extensive assortment of quality pH electrodes and sensors are available in a variety of body materials, body styles, fill types, reference styles, and junctions to meet the needs of your sample measurements.
We offer pH buffers, pH electrode storage solutions, electrode cleaning solutions, lab buffer kits, and electrode filling solutions for your pH measurement needs. Our pH buffers are NIST-traceable with certificates of analysis available for each solution batch.
Using high-quality solutions to calibrate and maintain your sensors is the best way to ensure your data is both accurate and reproducible. Learn more about why high-quality solutions deliver correct measurements every time.
Knowledge Base: Certificates of Analysis and SDS for Orion, ROSS, AQUAfast, AquaMate, and AquaSensors products.
Orion Meters Software and Upgrades
Thermo Scientific Orion benchtop and portable meters are designed to help deliver accurate, consistent results, day after day. Boost productivity by upgrading your software today.