The guide to selecting the right filtration product
Choosing the right filtration product can help reduce workflow delays, sample loss, contamination risk, and inconsistent downstream results. Beyond pore size and sample volume, factors such as sterility, membrane material, device format, handling steps, and filtration surface area can influence how efficiently a workflow performs. Thermo Fisher Scientific offers a broad filtration portfolio designed to help laboratories match the right product format to their application, from small-volume sample preparation and analytical filtration for QA/QC testing to larger-volume sterile filtration and cell preparation workflows.
What to consider when buying a filtration device
Sample volume is one of the first factors to consider when choosing a filtration device. Small-volume samples are often best suited to syringe filters or centrifugal devices, while larger volumes may require bottle-top filters, vacuum filtration units, or filtration manifolds. Matching the device format to the volume you need to process can help improve handling, reduce processing time, and minimise sample loss.
Pore size determines which particles or microorganisms are retained by the filter. Smaller pore sizes are typically used for fine particle removal or sterile filtration, while larger pore sizes may be suitable for clarification, prefiltration, or removal of larger debris. Selecting the correct pore size helps balance retention performance with flow rate and filter capacity.
The membrane material affects filtration performance, chemical compatibility, binding behavior, and suitability for different applications. Materials such as PES, nylon, PVDF, PTFE, cellulose acetate, and regenerated cellulose each offer different properties, making them better suited to specific sample types, solvents, proteins, or analytical workflows. Choosing the right membrane helps protect sample integrity and downstream results.
Sterility requirements depend on whether the filtered sample will be used in cell culture, microbiology, bioprocessing, or another sensitive workflow. Sterile filtration devices are designed to help reduce contamination risk when preparing media, buffers, reagents, or biological samples. For non-sterile applications such as general clarification or some analytical sample prep workflows, a non-sterile device may be sufficient.
Flow rate and throughput influence how quickly a sample can be processed and how efficiently the workflow can be completed. Higher-throughput applications may benefit from larger surface area filters, vacuum-driven devices, or manifold systems, while low-volume workflows may only require a syringe filter or small-format device. The right balance of membrane area, pore size, and device format can help improve filtration speed without compromising performance.
Chemical compatibility is important when filtering solvents, acids, bases, buffers, or other reagents that may interact with the membrane or device housing. Using an incompatible filter can affect performance, compromise the sample, or damage the device. Always match the membrane and housing material to the chemical properties of the sample and any downstream analytical requirements.
Frequently Asked Questions
What is the difference between clarification and sterile filtration?
Clarification removes visible particles, debris, precipitates, or larger contaminants from a sample. Sterile filtration is designed to remove microorganisms from compatible solutions. Thermo Scientific Nalgene membranes, syringe filters, and larger-format filter units can support clarification workflows, while Nalgene Rapid-Flow sterile disposable filter units and bottle-top filters are commonly used for sterile filtration of cell culture media, buffers, and other laboratory solutions.
What is the difference between a syringe filter and a bottle-top filter?
Thermo Scientific Nalgene syringe filters are generally used for small-volume sample preparation, such as clarifying samples before HPLC, UHPLC, or other analytical workflows. Thermo Scientific Nalgene Rapid-Flow bottle-top filters are designed for larger-volume filtration directly into a receiving bottle, making them suitable for preparing media, buffers, serum, additives, reagents, or other laboratory solutions.
When should I use a vacuum filtration unit?
Vacuum filtration units are useful when processing larger sample volumes or when faster filtration is needed. Thermo Scientific Nalgene Rapid-Flow sterile disposable filter units combine a filter and receiver system to support convenient filtration of media, buffers, and aqueous solutions. For cell preparation, the Thermo Scientific Nalgene Cell Strainer also offers optional vacuum support to help process difficult or slow-to-strain cell suspensions.
How do I choose a membrane material?
Membrane choice depends on the sample type, solvent, analyte, protein-binding requirements, and downstream application. Thermo Scientific Nalgene filtration products are available with membrane options such as PES, CN, SFCA, nylon, PTFE, PVDF, and cellulose-based materials across different formats, including bottle-top filters, filter units, syringe filters, and membranes. PES is often selected for aqueous filtration and low protein binding, nylon for general analytical sample preparation, PTFE for many organic solvents, and cellulose-based membranes for selected aqueous or biological workflows.
Why does chemical compatibility matter in filtration?
Chemical compatibility helps ensure that the filter membrane and housing can withstand the sample or solvent being filtered. An incompatible filter may swell, degrade, leach materials, or affect sample quality. Thermo Fisher Scientific offers filtration products in multiple membrane and housing material options, including Nalgene syringe filters and membranes, so users can select a format that matches their solvent, buffer, reagent, or analytical workflow requirements.
Can filtration affect sample recovery?
Yes. Sample recovery can be affected by membrane binding, hold-up volume, pore size, sample viscosity, and device format. Thermo Scientific Nalgene filtration products are available in multiple formats and membrane types to help users match the device to the sample and downstream workflow. For valuable biological samples, low-binding membrane options, appropriate pore size selection, and device formats with suitable sample capacity can help minimise sample loss.
What are analytical filters used for?
The notes describe typical analytical workflows: filtration, membrane transfer to dish, incubation, then colony counting or microscopy/particle examination. 2025 Training Analytical Filter…
Applications mentioned include water testing, pharma, beverages, hospitals (Legionella monitoring), and other industrial QC contexts.
For Research Use Only. Not for use in diagnostic procedures.