All TaqMan Real-Time PCR Master Mixes
For a comparison between our recommended TaqMan™ Master Mixes, please see Which TaqMan Master Mix Is Right for You.
Category | Product family | Description | Size |
---|---|---|---|
Gene expression | TaqMan™ Fast Advanced Master Mix | Highest performing master mix for gene expression applications | 5 mL pack |
TaqMan™ Universal Master Mix II | Ideal for customers working with multiple applications including SNP and CNV assays | 5 mL pack | |
TaqMan™ Gene Expression Master Mix | Precise quantification and sensitive detection down to one copy of target | 5 mL pack | |
TaqMan™ Universal PCR Master Mix | Ideal reagent solution when you need a Master Mix for multiple 5' nuclease DNA applications | 5 mL pack | |
TaqMan™ Fast Universal PCR Master Mix | Delivers results in 40 minutes for 40 cycles of PCR using the Applied Biosystems® 7900HT and Fast 7500 Fast Real-Time PCR Systems | 5-pack | |
Platinum™ Quantitative PCR SuperMix-UDG | Optimized to provide superior performance with either LUX™ Fluorogenic Primers or probe-based detection systems. | 500 reactions | |
EXPRESS qPCR Supermix Universal | Proven Invitrogen enzymes specifically tailored to deliver superior results with probe-based qPCR assays | 500 reactions | |
Genotyping | TaqPath™ ProAmp™ Master Mix* | Our highest-performing, high-throughput master mix for genotyping and copy number variation (CNV), including inhibited samples | 10 mL pack |
TaqMan™ GTXpress™ Master Mix | Delivers accurate genotyping results with robust performance in less than 50 min | 10 mL pack | |
TaqMan™ Genotyping Master Mix | Specifically formulated for detection of SNPs and insertions/deletions | 10 mL pack | |
TaqMan™ Sample-to-SNP™ Kit | Go from biological sample to genotyping results in less than one hour | 5 mL sample prep and 1 mL PCR | |
Custom master mixes | For plant samples › For other non-human sample types › |
||
PreAmp | TaqMan™ PreAmp Master Mix | Preamplifies limited cDNA without introducing amplification bias to the sample | 40 reactions |
* For laboratory use. General purpose reagent.