Find ELISA Kits by Target
Hundreds of targets, multiple species, several formats
Search, find, and compare over 2,000 ELISA kits by research target, species, synonyms, and gene symbol in an extensive portfolio of phosphorylation, neurobiology, cytokine, extracellular and intracellular targets.
Ready-to-use Coated ELISA Kits include reagents and components, such as pre-coated 96-well plates, detection antibodies, immunoassay standards, diluent buffers, wash buffers, stop solutions, and plate sealers. Uncoated ELISA Kits have added reagents needed to prepare and run the ELISA, including ELISA-optimized antibody matched pairs, standards, detection reagents, and coating buffers (plates are optional with 10 plate and 20 plate sizes).