Now you can quickly and easily order Invitrogen™ Stealth RNAi™ siRNA - predesigned RNAi sequences to the majority of human, rat, and mouse genes. Stealth RNAi siRNA sequences are synthesized using the same advanced chemistry as all Stealth RNAi siRNA, and pre-designed in silico using the BLOCK-iT RNAi Designer. In addition, an advanced mismatch alignment search results in Stealth RNAi siRNA sequences that essentially eliminate concerns of off-target effects. The new, easy-to-use Invitrogen™BLOCK-iT™ RNAi Express search engine allows you to quickly identify and order Stealth RNAi siRNA by keyword, accession number, pathway, or gene family.
Stealth RNAi pre-designs now on the siRNA selection tool!
Ready-to-order Stealth RNAi siRNA provides:
  • All of the advantages of Stealth RNA siRNA chemistry: reduced off-target and non-specific effects for clearer results
  • Three non-overlapping sequences supplied individually--experiments can be validated using independent sequences; ensures compatibility with publication requirements
  • Higher reduction of off-target effects--application of an advanced specificity search, in addition to BLAST
Get guaranteed results with Stealth RNAi [set of 3]
With Stealth RNAi siRNA the work of designing highly effective sequences has been done for you. The development of these molecules begins with identifying all human, rat, and mouse transcripts in the Unigene database. In four steps we select highly effective in silico-designed sequences (Figure 1).
Figure 1.  How Stealth RNAi siRNA sequences are designed.
Guarantee*: With the purchase of a Stealth RNAi siRNA Set, you will get guaranteed results that at least 2 out of the 3 sequences will result in at least 70% transcript knockdown, given that the transfection efficiency in your experiment is at least 80%.
Web Seminars and guides
Send your questions to our top researchers at