Lutetium • Lanthanide Rare Earth
Primary XPS region: Lu4d
Overlapping regions: N/A
Binding energies of common chemical states: N/A
Experimental information
N/A
Interpretation of XPS spectra
N/A
About this element
Element Crystal hex
Symbol: Lu
Date of discovery: 1907
Name origin: from Lutetia
Appearance: silvery
Discoverer: Georges Urbain
Obtained from: gadolinite, xenotime
Melting point: 1656 K
Boiling point: 3402 K
Density[kg/m3]: 9.85
Molar volume: 17.78 × 10-6 m3/mol
Protons/Electrons: 71
Neutrons: 104
Shell structure: 2,8,18,32,9,2
Electron configuration: [Xe]4f145d16s2
Oxidation state: 3
Crystal structure: hexagonal
Found with almost all other rare earth metals, but never by itself, lutetium is extremely difficult to isolate. Consequently, it is one of the most expensive metals, costing about six times as much per gram as gold. Not surprisingly, lutetium has no large-scale practical uses. Stable lutetium nuclides can be used as catalysts in cracking, alkylation, hydrogenation, and polymerization processes. It is used as a catalyst in refineries for petroleum cracking. Lutetium aluminum garnet has been proposed for use as a lens material in high refractive index immersion lithography.
element_lutetium
Products
Nexsa XPS
  • Módulo de basculación para mediciones de ARXPS
  • Fuente de iones de modo dual para capacidades de realización de perfiles de profundidad
  • Análisis de aislantes.
Hoja de estilo para tarjetas originales instrumentos
K-Alpha XPS
  • Espectroscopía de área seleccionable.
  • Monocromador microfocalizado.
  • Espectroscopía del estado químico de alta resolución.
ESCALAB Xi+ XPS
  • Espectroscopía de alta sensibilidad.
  • XPS con fuente de rayos X no monocromática.
  • Analizador hemisférico de 180°.
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Servicios de microscopía electrónica para
la ciencia de materiales
Para garantizar un rendimiento óptimo del sistema, le proporcionamos acceso a una red de expertos de primer nivel en servicios de campo, asistencia técnica y piezas de repuesto certificadas.