Xenon • Noble Gases
Primary XPS region: Xe3d
Overlapping regions: N/A
Binding energies of common chemical states:
Chemical stateBinding energy Xe3d5/2
Implanted Xe670 eV
Experimental information
N/A
Interpretation of XPS spectra
  • Xe3d region has well-separated spin-orbit components.
General comments
  • Xe is a gas and is not commonly found in XPS spectra.
  • Xe may be observed as implanted atoms, if a material has been subjected to Xe ion bombardment using Xe gas instead of the more usual Ar gas.
Element Crystal cub
About this element
Symbol: Xe
Date of discovery: 1898
Name origin: Greek meaning strange
Appearance: colorless
Discoverer: William Ramsay, Morris Travers
Obtained from: liquified air
Melting point: -111.9 K
Boiling point: -108.1 K
Density[kg/m3]: 5.894
Molar volume: 35.92 × 10-6 m3/mol
Protons/Electrons: 54
Neutrons: 77
Shell structure: 2,8,18,18,8
Electron configuration: [Kr]4d105s25p6
Oxidation state: 0,+1,+2,+4,+6,+8
Crystal structure: cubic
This odorless and colorless gas has a high molecular weight, which is often useful when used in atomic energy fields in bubble chambers, probes, and other applications. Xenon was identified a few weeks after the discovery of krypton and neon—all zero-valence elements know as noble gases. Xenon is used to make strobe lights. There are nine stable xenon isotopes, many of which have important medical applications. Three are used extensively in diagnostic procedures, while another is used in the treatment of prostate cancer.
Xenon • Noble Gases
Products
Nexsa XPS
  • Módulo de basculación para mediciones de ARXPS
  • Fuente de iones de modo dual para capacidades de realización de perfiles de profundidad
  • Análisis de aislantes.
Hoja de estilo para tarjetas originales instrumentos
K-Alpha XPS
  • Espectroscopía de área seleccionable.
  • Monocromador microfocalizado.
  • Espectroscopía del estado químico de alta resolución.
ESCALAB Xi+ XPS
  • Espectroscopía de alta sensibilidad.
  • XPS con fuente de rayos X no monocromática.
  • Analizador hemisférico de 180°.
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Servicios de microscopía electrónica para
la ciencia de materiales
Para garantizar un rendimiento óptimo del sistema, le proporcionamos acceso a una red de expertos de primer nivel en servicios de campo, asistencia técnica y piezas de repuesto certificadas.