Gold • Transition Metal
Primary XPS region: Au4f
Overlapping regions: N/A
Binding energies of common chemical states:
Chemical stateBinding energy Au4f7/2
Au metal84.0 eV
Experimental information
N/A
Interpretation of XPS spectra
  • Au4f region has well-separated spin-orbit components (Δ=3.7eV).
    • Peaks in the Au4f region have an asymmetric peak shape.
  • The Au4f7/2 peak can be a useful binding energy reference (Au4f7/2 at 84.00eV).
    • Binding energy shifts may be observed with Au nanoparticles, however.
      • Casaletto et al. analyzed Au nanoparticles on different metal oxide supports with XPS[1] and found two states of gold, Au0 and Au1+.
      • The Au1+ state was shifted by 2eV from the Au0 state.
Reference
[1] Casaletto et al., Surf. Interface Anal. 2006; 38: 215–218.
About this element
Element Crystal fcc
Symbol: Au
Date of discovery: circa 3000 BC
Name origin: Old English geolo
Appearance: gold
Discoverer: unknown
Obtained from: earth crust, copper ores
Melting point: 1337.58 K
Boiling point: 3080 K
Density[kg/m3]: 19300
Molar volume: 10.21 × 10-6 m3/mol
Protons/Electrons: 79
Neutrons: 118
Shell structure: 2,8,18,32,18,1
Electron configuration: [Xe]4f145d106s1
Oxidation state: 3,1
Crystal structure: cubic face centered
Gold, Latin aurium for "shining dawn", has been one of the most valued precious metals since prehistoric times and is mentioned several times in the Old Testament. Gold has been the standard for monetary exchange in many countries. While the metallic element takes on a yellow color en masse, when finely-divided it can also appear to be black, ruby, or purple in color. Being that gold is a soft metal, it is strengthened by being alloyed with other metals such as silver and copper. Gold’s heat and electrical conductivity qualities – as well as its resistance to heat, oxygen, moisture, and most corrosive agents – make it most applicable in coinage and jewelry. Seventy-five percent of all gold ever produced has been extracted since 1910. All gold ever refined would form a single cube twenty meters per side.
element_gold
Products
Nexsa XPS
  • Módulo de basculación para mediciones de ARXPS
  • Fuente de iones de modo dual para capacidades de realización de perfiles de profundidad
  • Análisis de aislantes.
Hoja de estilo para tarjetas originales instrumentos
K-Alpha XPS
  • Espectroscopía de área seleccionable.
  • Monocromador microfocalizado.
  • Espectroscopía del estado químico de alta resolución.
ESCALAB Xi+ XPS
  • Espectroscopía de alta sensibilidad.
  • XPS con fuente de rayos X no monocromática.
  • Analizador hemisférico de 180°.
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Servicios de microscopía electrónica para
la ciencia de materiales
Para garantizar un rendimiento óptimo del sistema, le proporcionamos acceso a una red de expertos de primer nivel en servicios de campo, asistencia técnica y piezas de repuesto certificadas.