Organism TypeHumanA T-lymphoma cell line established from a patient with Lennert's lymphoma. The cells show macrophage-dependent growth and macrophage-derived factors are able to replace macrophage functions.
TissuesLymphocyte
PhenotypeSuspension
Primaryno
Application