Search
Catalog Number(s)
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Inorganic Salts | |||
| Boric Acid (H3BO3) | 62.0 | 620.0 | 10.0 |
| Cobalt Chloride (CoCl2-6H2O) | 238.0 | 2.5 | 0.010504202 |
| Cupric sulfate (CuSO4-5H2O) | 250.0 | 2.5 | 0.01 |
| Ferric sulfate (FeSO4-7H2O) | 278.0 | 27.8 | 0.099999994 |
| Manganese Sulfate (MnSO4-H20) | 169.0 | 1690.0 | 10.0 |
| Potassium Iodide (KI) | 166.0 | 83.0 | 0.5 |
| Sodium Molybdate (Na2MoO4-2H20) | 243.0 | 25.0 | 0.10288066 |
| Zinc sulfate (ZnSO4-7H2O) | 305.0 | 860.0 | 2.819672 |
| Other Components | |||
| Sodium EDTA (Na2-EDTA) | 372.0 | 3730.0 | 10.026882 |
1. Murashige, T. and Skoog, F. (1962) Physiol. Plant. 15, 485.