Technical Resources - Media Formulations
Microvascular Growth Supplement (MVGS)
A sterile, concentrated (20X) solution intended for use with Medium 131 to culture human microvascular endothelial cells. Below is the composition list. The concentrations are proprietary.
ComponentsMolecular WeightConcentration (mg/L)mM
Proteins
Basic FGFconfidentialn/a
EGF (human recombinant)confidentialn/a
Other Components
FBSconfidentialn/a
Heparinconfidentialn/a
Hydrocortisoneconfidentialn/a
dbcAMPconfidentialn/a
Reference: