Search
Having difficulties with your Dynabeads cell isolation and expansion experiments?
We are dedicated to your success. Get back on track. View our expert recommendations for commonly encountered problem scenarios related to your Dynabeads cell isolation and expansion experiments.
View the relevant questions below.
Beginning your experiment?
Visit our
Here are some recommendations to improve target cell purity when using Dynabeads magnetic beads for negative isolation:
Here are some recommendations to improve final recovery/yield when using Dynabeads magnetic beads for negative isolation:
Platelet activation often constitutes a problem when isolating monocytes, as activated platelets bind to monocytes and produce unfavorable aggregates of monocytes and platelets. There are several factors that can trigger platelet activation, such as temperature changes, mechanical factors (e.g., shear), exposure to the anticoagulant heparin (often present in sampling tubes), etc. In order to avoid platelet activation (and subsequent monocyte aggregation), there are a few general recommendations that should be followed:
Yes, the cloudy appearance is normal for DETACHaBEAD™ beads because the protein content is very high and the protein can precipitate. This is not bacterial contamination and does not affect the recovery or viability of the isolated and released cells. Mix the solution thoroughly before use.
Here are some recommendations:
Here are some suggestions to improve cell yield when isolating cells with any of the Dynabeads positive isolation kits:
Here are some suggestions:
The best choice is to use the Dynabeads™ FlowComp™ Flexi Kit in combination with a suitable target antibody for cell isolation. The Flexi kit contains both a DSB-X biotinylation kit for biotinylation of your antibody, the modified streptavidin-coated FlowComp™ Dynabeads™ magnetic beads, and the FlowComp™ Release Buffer required for performing isolation and release.
Here are some suggestions:
Here are some suggestions:
Here are a few tips to keep the optimal cell culture growth conditions:
Have input on this page? Use the "Give Feedback" button on the right.
Need more information? Contact us ›
For Research Use Only. Not for use in diagnostic procedures.