Chromatography resins for therapeutic manufacturing
Scientists need purification solutions that maintain performance from development through cGMP manufacturing. Chromatography plays a role in achieving consistent purity, yield, viral clearance, and productivity targets as processes evolve from small-scale research to commercial production. Thermo Fisher Scientific supports this transition with chromatography solutions designed to optimize recovery, reduce impurities, and maintain throughput across diverse purification workflows.
Affinity chromatography resins
Affinity resin allows capture with high purity and yield in one step. While often used for antibodies, CaptureSelect technology also supports bispecific antibodies, fragments, Fc-fusion proteins, recombinant proteins, and viral vectors using mild elution conditions.
Ion exchange chromatography resins
Ion exchange chromatography (IEX) separates biomolecules through charge interactions with functional groups. These resins are available in both strong and weak base or acid chemistries, suitable for bind/elute or flow-through purification of antibodies, vaccines, viral vectors, and other biologics.
Anion exchange chromatography resins
Anion exchange chromatography (AEX) resins separate biomolecules based on charge, capturing negatively charged species through interactions with positively charged groups. Strong AEX resins maintain capacity across a wide pH and conductivity range for both capture and polishing.
Cation exchange chromatography resins
Cation exchange chromatography (CEX) resins separate positively charged biomolecules using negatively charged functional groups on the resin matrix. This approach is often used to purify proteins, peptides, enzymes, and other biologics during the production of therapeutic molecules. POROS CEX resins offer high capacity, chemical stability, and consistent performance from process development to commercial manufacturing.
Mixed-mode chromatography resin
Mixed-mode chromatography uses ion exchange and hydrophobic interactions within a single chemistry to improve selectivity in complex samples. POROS mixed-mode cation-exchange resin is designed for flow-through polishing and is used to reduce aggregates, host cell proteins, and other impurities while supporting high product recovery and scalability from process development through cGMP.
Protein A chromatography resin
Protein A (ProA) chromatography is often used for antibody purification. MabCaptureC resin offers high binding capacity, alkaline stability, and scalability, while keeping low cost of goods in mind, making it suitable for process development through cGMP manufacturing.
Hydrophobic interaction chromatography resins
Hydrophobic interaction chromatography (HIC) separates biomolecules by surface hydrophobicity. HIC is commonly applied as a polishing step, improving purification where charge-based methods may be limited.
Supporting diverse modalities and applications
ADCs
Antibody-Drug Conjugates (ADCs) workflows rely on chromatography steps to separate complex conjugates and reduce impurities. These approaches support consistent production of ADCs for therapeutic research and biomanufacturing.
Cell Therapy
Cell therapies utilize living cells as treatments, necessitating strict control over inputs and processing steps. Chromatography is applied to prepare ancillary materials, such as cytokines or viral vectors, that are used during cell manufacturing.
Gene Therapy
Gene therapy often relies on viral vectors, such as adeno-associated virus (AAV) or lentivirus, to facilitate the transfer of genetic material. Chromatography separates drug product from product- and process-related impurities in both capture and polishing steps.
mAbs
Monoclonal antibody purification typically starts with Protein A chromatography to capture the antibody from cell culture fluid. Additional steps, such as ion exchange, are used to help reduce aggregates, variants, or process-related impurities.
mRNA
Chromatography in mRNA production separates full-length transcripts from double-stranded RNA and truncated products. The purified RNA can then be used in formulation or downstream steps, such as lipid nanoparticle assembly.
Vaccines
Chromatography is often used in vaccine manufacturing for both the capture and polishing of the vaccine drug product. The choice of method depends on the platform, for example, protein subunits, viral vectors, or nucleic acid vaccines.
Enabling purity and yield in advanced applications
Plasma fractionation
Plasma fractionation is used to separate plasma proteins, such as albumin, immunoglobulins, and clotting factors, for therapeutic applications. Chromatography steps integrated into these workflows are designed to enhance selectivity and consistency to improve the overall yield of precious plasma-derived products. Thermo Fisher Scientific supports improved productivity in plasma purification processes with affinity and polishing resins implemented at various stages.
Choose the right resins for your workflow
Frequently asked questions
Chromatography resins are stationary phase matrices used in purification to bind, separate, or polish biomolecules such as proteins, antibodies, and nucleic acids. Different resin chemistries—including affinity, ion exchange, hydrophobic interaction, and mixed-mode—are selected based on the properties of the molecules and the process goals.
Chromatography resins are generally classified by their mode of interaction with biomolecules. Common types include affinity resins, IEX resins, HIC resins, and mixed-mode resins.
POROS resins offer high binding capacity, mechanical strength, and rapid mass transfer, supporting efficient purification at laboratory and manufacturing scales. CaptureSelect resins offer engineered ligand specificity for improved selectivity and product yield in manufacturing complex biologics, including antibody fragments and viral vectors. These technologies facilitate consistent performance and scalable bioprocessing.
Chromatography resins are produced from agarose, silica, or polymer backbones that are functionalized with specific ligands. The particle size, porosity, and surface chemistry are precisely controlled during manufacturing to achieve consistent binding capacity, chemical stability, and flow performance across purification scales.
Thermo Fisher Scientific offers custom resin development services for projects that require specific ligand chemistries or performance characteristics. Options include custom ligand and resin design, scale-up manufacturing, and support to meet regulatory or process requirements. Learn more about custom resin development or custom POROS development.
Resins are often used with other chromatography products that refine purification or track process stability. Process buffers are fundamental to control pH and conductivity. Scientists may also require analytical chromatography systems to verify the quality of separation and ligand conjugates for further analytical development. In practice, combining these elements helps maintain consistency across purification runs.
Researchers can request free chromatography resin samples to test performance before scale-up. Samples are available through the Biotherapeutic Purification Sample Form.
Explore chromatography techniques
Chromatographic techniques—such as affinity, ion-exchange, hydrophobic interaction, and mixed-mode chromatography—are used at different stages of purification. Each mode applies distinct binding principles, making it important to match the technique with your target molecule and process needs.
Discover other chromatography products
Thermo Fisher Scientific offers a broad chromatography portfolio that supports purification from early research to commercial-scale manufacturing. Each product is designed to fit specific workflow needs across a range of modalities, including mAbs, therapeutic proteins, cell and gene therapies, mRNA, ADCs, and vaccines. Together, these chromatography products enable consistent performance, scalability, and flexibility in bioprocessing applications.
CaptureSelect and POROS Chromatography
CaptureSelect and POROS resins support the purification of antibodies, viral vectors, and recombinant proteins. CaptureSelect resins offer selectivity and gentle elution, while POROS resins offer speed and scalability for consistent large-scale chromatography performance.
Chromatography columns
Pre-packed chromatography columns are designed to simplify method setup and reduce preparation time. Available in multiple sizes, they support capture, intermediate, and polishing steps, enabling reliable data for process development and scale-up.
Membrane chromatography
Membrane chromatography enables rapid purification with low backpressure. These high-throughput formats help reduce processing time and are well-suited for biologics production, from process development through manufacturing.
96-well screening plates
The 96-well screening plate format enables rapid resin evaluation by allowing multiple purifications to run in parallel. This approach facilitates the comparison of binding and wash conditions at a small scale, supporting the development of efficient methods before larger-scale purification studies.
Find the right resins for your workflow
Selecting the right chromatography resins for your workflow depends on the molecule type, process objectives, and desired purity. Compare chemistries, binding capacities, and scalability data in one place.
Chromatography resources
Explore protocols, application notes, and troubleshooting for chromatography techniques. You’ll also find videos, webinars, and how-to articles on column packing, method development, and scale-up, plus calculators and selection tools to support process decisions.
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.