On this page:
Designed as a cost-effective mixing solution
The Thermo Scientific HyPerforma Mixtainer System is an integrated single-use sterile unit designed for optimal mixing of cell culture media, process liquids, buffers, reagents, and bulk product. The HyPerforma Mixtainer System is designed as an alternative to conventional bag technology and mixing in open-tank liners. It can hold, store, stir, and suspend sterile fluids in an enclosed environment. Encompassing a single-use Thermo Scientific HyPerforma Mixtainer BioProcess Container (BPC) equipped with an integrated magnetic stir plate, a supporting conical drum, and a dolly to align the components help enable easy mobility, the Mixtainer System provides an ideal solution for your bioprocess workflow.
Key features
  • Reliable BPC constructed of Thermo Scientific CX5-14 film, with a plastic dish welded to the base
  • Welded plastic dish containing a magnetic stir bar, held in the proper operating position using an integrated locking ring
  • Available in 50, 100, and 200 L unit volumes
  • BPCs available for liquid-to-liquid and powder-to-liquid mixing
  • Optimized supporting hardware allowing for mixing, storage, transport, and discharge in a closed system to help minimize risk of cross-contamination
Areas of application
Our HyPerforma Mixtainer System can be used in many applications, such as media and buffer hold and transportation in GMP and development environments:
  • Cell culture
  • Filtration
  • Chromatography
  • Formulation
HyPerforma Mixtainer BPC options
Four ports, top dispense
  • Line 1: 9.5 mm (0.38 in.) quick-connect insert with 16.5 cm (6.5 in.) dip tube; tube length: 91 cm (36 in.)
  • Line 2: 6.3 mm (0.25 in.) Luer-lock insert; tube length: 91 cm (36 in.)
  • Line 3: 6.3 mm (0.25 in.) Luer-lock body; tube length: 91 cm (36 in.)
  • Line 4: 9.5 mm (0.38 in.) quick-connect body; tube length: 91 cm (36 in.)
Need to contact a specialist or get support for your single-use mixer?
Single-Use Bioprocessing Equipment and Automation
We are committed to delivering exceptional performance, reliability, and quality.
Single-Use Bioprocessing
Our innovative single-use technologies enable you to accelerate biological manufacturing processes to create flexibility and operate more dynamically within upstream and downstream workflows.
BioProcessing Containers and Nalgene Bottles
We offer high-quality storage, transport, and sampling solutions, including Thermo Scientific BioProcess Containers (BPCs) and Thermo Scientific Nalgene containers.
Speak to a specialist
Connect with a Bioprocessing Single-Use Technology representative.
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.