Search
Raman spectroscopy provides many advantages for users:
The MarqMetrix All-In-One Process Raman Analyzer is specifically designed to integrate the power and sensitivity of Raman spectroscopy directly into workflows to generate insights for process monitoring and process control.
That makes the MarqMetrix All-In-One analyzer ideal for process optimization across many industries and applications including biotechnology, bioprocessing, pharmaceuticals, biopharmaceuticals, oil and gas, polymers, chemicals, sustainable fuels, hydrogen, carbon capture, food and beverage, industrial fermentation, process integration, and more.
With the MarqMetrix All-In-One Process Raman analyzer, biopharma and pharma companies can employ the benefits of Raman spectroscopy for development, optimization and process control. The analyzer provides real-time compositional analysis with simple installation, straightforward control, high sensitivity, reproducibility and reliability from development bench to continuous process analysis.
Biopharma operations can employ the MarqMetrix All-In-One analyzer as a process analytical technology (PAT) to optimize both upstream and downstream processes. Applications include integration with bioreactors for real-time metabolite monitoring, automated feedback for glucose concentrations, efficiencies in alternative protein analysis such as cultivated meat along with demonstrated chemometric transferability.
The MarqMetrix All-In-One analyzer supports pharma applications from drug discovery through wet granulation and drying to continuous flow reactor monitoring and control. In-line integration with hot-melt extrusion processes allows users to manage temperature, pressure, and feed rate for enhanced blend uniformity and product quality. The analyzer also enables verification of raw materials and active pharmaceutical compounds and can be used in quality testing and assurance – even through transparent packaging.
Optimization of manufacturing processes becomes increasingly important as the polymer sector pushes the technical frontiers of lighter chemical composites and tougher materials.
The MarqMetrix All-In-One Process Raman Analyzer can be used to analyze the composition and structure of polymers during production and for final product quality control. Specialized probes are designed for high pressures, extreme temperatures, and even corrosive chemicals to enable measurements throughout polymer production. The analyzer helps users quickly describe composition, determine structure, and optimize molecular weight in both solid and liquid forms.
Accuracy is more important than ever in the energy and chemical sectors. Understanding the chemical composition of raw materials and ensuring the uniformity of finished products can provide a significant competitive edge at all stages of the process.
The MarqMetrix All-In-One Process Raman Analyzer is a solid-state system that can accelerate analysis, provide flexibility by measuring liquids and gases, and provide data correlated to American Society for Testing and Materials (ASTM), Gas Producers of America (GPA), and American Gas Association (AGA) standards. The analyzer and accompanying sampling optics have been successfully used in midstream and downstream applications such as:
Raman field analysis is an effective technique for determining the chemical composition of natural gas in the midstream phase. The MarqMetrix All-In-One Process Raman Analyzer can be easily carried on-site to swiftly assess C1-C6+, CO2, H2, and N2, providing real-time insights for confident decision-making.
Refined fuel qualities such as API, benzene levels, RON, and MON must be assessed accurately and precisely in the downstream segment to allow refineries to expedite quality assurance and provide fuels to market fast. The MarqMetrix All-In-One Process Raman Analyzer can quickly and accurately test several properties of refined fuels, which streamlines an on-site lab's analysis procedure and speeds up quality assurance.
Producers of sustainable fuels can also benefit from process Raman spectroscopy. The MarqMetrix All-In-One analyzer can be used in processes for quality control and monitoring, contaminant detection, carbon footprint assessment, and research and development.
Hydrogen is increasingly being looked to as an option to help create a decarbonized energy future. Trustworthy transportation and storage of liquid hydrogen is pivotal for its widespread use. Ensuring safety and efficiency in hydrogen liquefaction and storage is essential, and the MarqMetrix All-In-One Process Raman Analyzer can help companies optimize and monitor those processes.
Carbon dioxide gathered during industrial processes is converted into common items such as gasoline, chemicals, and rubber. To achieve the required yield and control product quality, the carbon capture and any associated fermentation processes must be continuously monitored. The MarqMetrix All-In-One Process Raman Analyzer analyzes the feed stream, analytes (nutrients), byproducts, and products in real time.
Lithium-ion battery development encompasses multiple stages, from material synthesis to cell assembly and recycling. The stages may even happen at different facilities, particularly with recycling of spent batteries. Precise monitoring is required at each stage to ensure quality, efficiency, and safety. The MarqMetrix All-In-One Process Raman Analyzer provides rapid insights across many stages in the battery workflow.
The MarqMetrix All-In-One Process Raman Analyzer enables users to achieve accurate and dependable compositional measurement, supporting the production of complex flavors and textures. Businesses benefit from the analyzer’s real-time composition characterization to assist in optimizing flavor profiles and physical product properties.