Search
Simplify setup and ensure reliable connections with pre-assembled hose, fittings, clamps, and insulation kits designed for ease of installation and maintenance. Ideal for both laboratory benches and industrial installations.
Explore the various models of TSC Series Recirculating Chillers to find the perfect fit for your application. Each model offers unique features and capabilities to ensure optimal performance.
Choose from a range of recirculating chillers. It's as easy as 1-2-3-4.
Small capacity chillers |
Mid-capacity chillers |
Large capacity chillers |
||||
TSC08 |
TSC16 | TSC35 | TSC66 | TSC90 | TSC210 |
TSC260 |
Compact, energy-efficient chiller with low-GWP refrigerants and precise thermal control for benchtop instruments and small heat-load applications |
Versatile small-capacity chiller delivering efficient, low-GWP cooling performance and stable temperature control for a wide range of laboratory instruments |
High-stability chiller providing improved efficiency and advanced refrigeration control for higher-demand laboratory and light industrial cooling needs. |
Reliable, mid-capacity chiller with advanced thermal controls and low-GWP refrigerants for analytical platforms and process cooling applications |
Powerful, energy-efficient chiller offering precise cooling and enhanced system stability for high-performance thermal management workflows. |
Industrial-grade chiller delivering exceptional cooling capacity, advanced compressor technology, and optimized thermal stability for large pilot systems and process-intensive applications |
Heavy-duty, high-capacity chiller engineered for demanding industrial processes, featuring low-GWP refrigerants and enhanced performance control. |
| *Cooling capacity @ 20°C setpoint | ||||||
60 Hz: 800W |
60 Hz: 1600W |
60 Hz: 3500W |
60 Hz: 6600W |
60 Hz: 9000W |
60 Hz: 21000W |
60 Hz: 26000W |
Condenser options |
||||||
A: Air-cooled condenser |
A: Air-cooled condenser |
A: Air-cooled condenser |
A: Air-cooled condenser |
A: Air-cooled condenser |
A: Air-cooled condenser |
A: Air-cooled condenser |
| Process fluid setpoint temperature range | ||||||
Standard range models: |
Standard range models:
|
Standard range models: |
Standard range models: +5°C to +40°C (+41°F to +104°F) Extended range models: -5°C to +90°C +23°F to +194°F) |
|||
| Available pump options | ||||||
T1, turbine |
T1, turbine |
T1, turbine |
T2, turbine |
T2, turbine |
T3, turbine |
T3, turbine |
| Available electrical options | ||||||
B: 115V/60Hz or 100V/50-60Hz 208-230V/60Hz/1 or 220V/50Hz/1 |
B: 115V/60Hz or 100V/50-60Hz 208-230V/60Hz/1 or 220V/50Hz/1 |
F: 208-230V/60Hz/1 or 220V / 50Hz/1 |
F: 208-230V/60Hz/1 or 220V/50Hz/1 |
G: 208-230V/60Hz/3 or 200-220V/50Hz/3 |
G: 208-230V/60Hz/3 or 200-220V/50Hz/3 460V/60Hz/3 or 400V/50Hz/3 |
G: 208-230V/60Hz/3 or 200-220V/50Hz/3 460V/60Hz/3 or 400V/50Hz/3 |
| Applications | ||||||
Analytical instruments, Bench-top reactors, Lasers and optics, Life science workflows |
Analytical instruments, Bench-top reactors, Lasers and optics, Life science workflows |
Analytical instruments, Bench-top reactors, Lasers and optics, Life science workflows |
Chemical / Pharmaceutical Pilot Systems, Laser and Semiconductor Equipment, Food & beverage Pilot Plants, Thermal cycling and stability testing for electronics |
Chemical / Pharmaceutical Pilot Systems, Laser and Semiconductor Equipment, Food & beverage Pilot Plants, Thermal cycling and stability testing for electronics |
Pilot-scale or process-intensive cooling, Industrial/Semi-conductor manufacturing tools. Large Reactors |
Heavy-duty heat loads, industrial process cooling, Pharmaceutical & Biotech Production |
| Ordering information | ||||||
| Standard | ||||||
*Cooling capacity based on no pump back pressure. Heat input from the pump will result in a reduction in cooling capacity. The cooling capacity reduction will vary.
Other application parameters, fluids, fluid temperatures, ambient temperatures, altitude or operating voltages will affect performance. Refer to owner's manual
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.