Forensic ready designation
Consumables and supplies fit for the rigors of the most sensitive PCR-based science
As the importance of human forensic science grows, so does the need for increased scientific rigor in these studies. Our Forensic ready product designation indicates products that meet the high standards necessary for rigorous scientific study and reliable results. The Thermo Scientific Forensic ready designation includes combination testing for sterility and elimination of endotoxins (see Sterile designation), as well as testing for the elimination of RNase, DNase, human DNA, and PCR inhibitors (see PCR Ready designation).  Products that meet this dual designation support the demands of Forensic science where sensitive Polymerase Chain Reaction (PCR) methods are used, and where sample material is limited and valuable.  
To receive the Forensic ready designation, products must meet the following criteria:
 Minimum Sterility Assurance Level (SAL) of 10-6
 Endotoxin (pyrogen) free
✓ Human DNA free
 RNase Free
 DNase Free
 PCR inhibitor free
Additional criteria met:
✓ Made from high quality resins and raw materials
✓ Manufactured in accordance with ISO 9001 or 13485 standards
✓ Low wetting for accurate liquid measurements
✓ High chemical resistance for durability in most routine lab work
✓ High thermal resistance for stability under wide range of temperatures
✓ High centrifugation stability*
✓ High transparency for good visual sample monitoring*
✓ Precisely shaped for outstanding function and critical measurements
*For applicable products only
Shop Forensic Ready Products
Thermo Scientific Forensic ready designated products are recommended for use with the following methods and application areas:
Application areas
→ Forensic human identification
→ Molecular biology
Techniques
→ DNA analysis (PCR, restriction analysis, hybridization, sequencing, NGS)
→ Isolation and storage of RNA
→ Isolation and storage of DNA
→ Mitochondrial DNA analysis
→ RNA analysis
Browse popular products certified under our Thermo Scientific Forensic ready designation. 
ART System Tips
ClipTip Tips
HPT-Softfit-L Hinged Rack