Search
Experience the first-ever chemically defined baculovirus expression system that can generate three times more protein compared to existing insect expression platforms. The ExpiSf Expression System enables greater consistency across multiple expression runs, all in less time. Put the newest member of the Expi system family to the test in your lab, from discovery to production.
Share your protein expression application needs with us, and one of our protein expression specialists will contact you to discuss the solutions we can provide.
“The new ExpiSf Expression System offers a convenient virus generation and protein expression cell line in only two steps, for both secreted and intracellular proteins,we predict that using the ExpiSf system will save us about 30 percent in the time taken for protein expression and make cell maintenance simpler.” Xuan Droz, Ph.D., project leader, Redbiotec AG
Undefined insect cell media components like yeastolate make it very difficult to get consistent performance. The Gibco ExpiSf system is the first ever baculovirus expression system that includes both a chemically defined medium and protein expression enhancer for reliable results you can count on. This insect cell expression system also utilizes a non-engineered derivative of Sf9 insect cells that have been adapted for high-density suspension growth and a specialized vector for insect cells.
*not included in China
** not included in all regions
The ExpiSf Expression System is designed to deliver consistent cell growth and protein expression, run after run. With our new Sf9 expression system, inconsistent results are a thing of the past. The superior performance of the chemically defined ExpiSf system delivers consistent results across expression runs and over multiple cell passages. One reliable system gives you one less variable to worry about.
It’s revolutionary—the ExpiSf system outperforms existing Sf9 expression platforms with 3x higher protein yield. The ExpiSf Expression System achieves protein yields that are vastly superior to traditional insect platforms. In performance tests against other suppliers’ media, the ExpiSf system demonstrated greater protein expression, delivering 3x more protein.
Figure 1. Recombinant protein titers in ExpiSf and other baculovirus expression systems. Expression levels of green fluorescent protein (GFP), Fc fusion protein, and tumor necrosis factor alpha (TNF-a) in ExpiSf and other baculovirus-based insect cell protein expression products are shown. ExpiSf protein titers are >3, >4, or >5 times those of other supplier’s insect products, respectively.
Figure 2. Expression of three different protein categories achieve expression levels >3x higher in the ExpiSf Expression System. Comparative western blotting analysis of a membrane protein (top panel), a receptor-type protein kinase (middle panel) and a tyrosine-protein kinase (bottom panel) produced in ExpiSf and control (Sf9 cells in Sf-900 II SFM) is shown. P0, P1, and P2 signify non-amplified (P0) or amplified (P1, P2) baculovirus stock used for the respective groups. *1.25x104 total cells/lane, **2.5x104 total cells/lane.
| Thermo Fisher | Thermo Fisher | Thermo Fisher | Expression Systems | MilliporeSigma | Oxford Expression Technologies | HyClone | |
|---|---|---|---|---|---|---|---|
| Cells | ExpiSf9 | Sf9 | Sf9 | Sf9 | Sf9 | Sf9 | Sf9 |
| Medium | ExpiSf CD | Sf-900 II SFM | Sf-900 III SFM | ESF 921™ | EX-CELL™ 420 | ESF 921™ | SFX-Insect™ |
| Transfection Reagent | ExpiFectamine Sf | Cellfectin II | Cellfectin II | Expres2 TR | Insect GeneJuice™ | Expres2 TR | Cellfectin II |
| Baculovirus System | Bac-to-Bac | Bac-to-Bac | Bac-to-Bac | BestBac™ | BacMagic™ | flashBAC™ | Bac-to-Bac |
| Cost of 1L expression | $334 | $235 | $260 | $186 | $332 | $110 | $243 |
| Culture volume (mL) for equivalent yield* | 300 | 1200 | 1700 | 3100 | 1700 | 3100 | 1500 |
| Number of 2L shake flasks needed | 1 | 2 | 3 | 6 | 3 | 6 | 3 |
| Total cost for same amount of protein | $112 | $294 | $454 | $579 | $563 | $343 | $368 |
| Savings | $65 | $226 | $351 | $335 | $115 | $139 |
System components work synergistically for maximum performance. Other suppliers offer separate reagents for protein production using the baculovirus expression vector system, but there is only one system that’s fully optimized with a streamlined workflow. The ExpiSf system contains integrated components that work together to deliver maximum protein yields.
The ExpiSf Expression System delivers protein in half the time. Bacmid DNA transfection in suspension using the ExpiSf system allows for efficient baculovirus generation without the need for virus amplification. That means you can get your protein in as few as six days.
The ExpiSf system accelerates development from small to large scale. From deep-well plates to shake flasks, the ExpiSf system delivers the same high levels of volumetric yield, making the system excellent for both discovering new therapies and producing them in large quantities. We manufacture ExpiSf CD Medium in cGMP-compliant facilities and offer custom packaging to scale with your development, so you can be confident that our product quality enhances yours.
We offer a full array of innovative products and services, including fast and flexible cell line development, media and feeds, class-setting technologies, quality custom media development, and media optimizing services.
Learn more about how we can customize your media formulation and format
Protein Expression Learning Center
Access protein expression educational resources for better experiment planning and execution.
Protein Expression Support Center
Find technical support recommendations for your protein expression workflows, including tips for experimental setup and in-depth troubleshooting help.
Protein Expression Publication Hub
Access a curated list of publications that highlights the advantages of our Expi protein expression systems.