Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| L-Arginine hydrochloride | 211.0 | 126.64 | 0.60018957 |
| L-Cystine 2HCl | 313.0 | 31.0 | 0.09904154 |
| L-Glutamine | 146.0 | 292.0 | 2.0 |
| L-Histidine hydrochloride-H2O | 210.0 | 42.0 | 0.2 |
| L-Isoleucine | 131.0 | 52.0 | 0.39694658 |
| L-Leucine | 131.0 | 52.0 | 0.39694658 |
| L-Lysine hydrochloride | 183.0 | 72.5 | 0.39617488 |
| L-Methionine | 149.0 | 15.0 | 0.10067114 |
| L-Phenylalanine | 165.0 | 32.0 | 0.19393939 |
| L-Threonine | 119.0 | 48.0 | 0.40336135 |
| L-Tryptophan | 204.0 | 10.0 | 0.04901961 |
| L-Tyrosine disodium salt | 225.0 | 52.0 | 0.23111111 |
| L-Valine | 117.0 | 46.0 | 0.3931624 |
| Vitamins | |||
| Choline chloride | 140.0 | 1.0 | 0.007142857 |
| D-Calcium pantothenate | 477.0 | 1.0 | 0.002096436 |
| Folic Acid | 441.0 | 1.0 | 0.0022675737 |
| Niacinamide | 122.0 | 1.0 | 0.008196721 |
| Pyridoxal hydrochloride | 204.0 | 1.0 | 0.004901961 |
| Riboflavin | 376.0 | 0.1 | 2.6595744E-4 |
| Thiamine hydrochloride | 337.0 | 1.0 | 0.002967359 |
| i-Inositol | 180.0 | 2.0 | 0.011111111 |
| Inorganic Salts | |||
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 140.0 | 1.2612612 |
| Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 97.67 | 0.8139166 |
| Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
| Potassium Phosphate monobasic (KH2PO4) | 136.0 | 60.0 | 0.44117647 |
| Sodium Chloride (NaCl) | 58.0 | 7500.0 | 129.31035 |
| Sodium Phosphate dibasic (Na2HPO4) anhydrous | 142.0 | 48.0 | 0.33802816 |
| Other Components | |||
| D-Glucose (Dextrose) | 180.0 | 1000.0 | 5.5555553 |
| HEPES | 238.0 | 5958.0 | 25.033613 |
| Phenol Red | 376.4 | 10.0 | 0.026567481 |
1. Eagle, H. (1959) Science 130:432.
2. Lolait, S.J., Harmer, J.H., Dahl, D., et al., (1983) Aust. J. Exp. Biol. Med. Sci., 61:439. (rat astrocytes).
3. Norris, S.J. and Edmondson, D.G. (1986) Sex. Transm. Dis., 13:207 (Rabbit epithelial cells).
21575 - MEM, Hank's
Cat No:
21575022