Search
Contains no insulin.
B-27 Serum-Free Supplement is used for growth and long-term viability of hippocampal neurons. This supplement is to be used with a combination of basal medium such as Neurobasal Medium. Below is the composition list. The concentrations are proprietary.
Catalog Number(s)
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Vitamins | |||
| Biotin | confidential | n/a | |
| DL Alpha Tocopherol Acetate | confidential | n/a | |
| DL Alpha-Tocopherol | confidential | n/a | |
| Vitamin A (acetate) | confidential | n/a | |
| Proteins | |||
| BSA, fatty acid free Fraction V | confidential | n/a | |
| Catalase | confidential | n/a | |
| Human Transferrin | confidential | n/a | |
| Superoxide Dismutase | confidential | n/a | |
| Other Components | |||
| Corticosterone | confidential | n/a | |
| D-Galactose | confidential | n/a | |
| Ethanolamine HCl | confidential | n/a | |
| Glutathione (reduced) | confidential | n/a | |
| L-Carnitine HCl | confidential | n/a | |
| Linoleic Acid | confidential | n/a | |
| Linolenic Acid | confidential | n/a | |
| Progesterone | confidential | n/a | |
| Putrescine 2HCl | confidential | n/a | |
| Sodium Selenite | confidential | n/a | |
| T3 (triodo-I-thyronine) | confidential | n/a | |
B-27 Serum-Free Supplement (50X) liquid
Cat No:
17504001
,
17504044
B-27 Supplement Minus AO (50X) liquid
Cat No:
10889038
B-27 Supplement Minus Vitamin A (50X) liquid
Cat No:
12587001
,
12587010