Human HEK 293 cells loaded with FluoVolt™ Membrane Potential Probe
Cells were imaged with 10 millisecond illumination pulses and images acquired with 2X binning. Traces show fluorogenic responses as cells are depolarized (A) at 2 second intervals from -100 mV to +30 mV or (B) in single steps from -80 mV to 0 mV at 2 second intervals.
Cells were imaged with 10 millisecond illumination pulses and images acquired with 2X binning.  Traces show fluorogenic responses as cells are depolarized (A) at 2 second intervals from -100 mV to +30 mV or (B) in single steps from -80 mV to 0 mV at 2 second intervals.
Related Products
Related Images