Search
Wednesday, July 10th
Title: Innovations in Pluripotent Stem Cell Culture
Speaker: Sara Pérez Muñoz, PhD Field Applications Scientist with Thermo Fisher Scientific
Sara will discuss Thermo Fisher Scientific's commitment to reliable and high-quality cell culture solutions through the latest developments in pluripotent stem cell applications
Thursday, July 11th—Lunchtime, hors d'oeuvres provided
Title: TBD
Speaker: TBD
Learn about our latest instruments, products, and services that support your stem cell workflow.
Your source for recombinant proteins including growth factors and cytokines with over 35 years of research and manufacturing experience.
TBD
The Gibco CTS Rotea Counterflow Centrifugation System is a closed, cell-processing system for cell therapy that offers exceptional cell recovery, flexible input and output volume capability, and high-throughput rate. The compact footprint, process flexibility, and sterile single-use kits allow the system to seamlessly scale with your process, from research, through clinical manufacturing.
The Gibco CTS Xenon Electroporation System is a closed, scalable, electroporation system for GMP-compliant cell therapy manufacturing. It offers reliable delivery of DNA, RNA, proteins, and other molecules into cells, with exceptional cell viability and recovery, leading to efficient transfection and genome editing, even in hard-to-transfect cells. Offering high performance in volumes of 1 mL or 5–25 mL, the CTS Xenon system can be used to seamlessly scale your cell therapy workflow from process development through clinical manufacturing.
The Neon NxT Electroporation System is an electroporation machine that uses an electronic pipette tip as an electroporation chamber. The design and performance of the electronic pipette transfection chamber result in increased cell viability and transfection efficiency compared to traditional cuvette-based electroporation systems.
The proprietary tip technology of Neon NxT Electroporation System enables scientists to reduce sample transfer loss while simplifying their workflow. The system was designed for electroporation of mammalian cells, but some customers have found it to be successful for other cell types such as insect cell cultures and parasites.
The advantages of this electroporation device are:
The design of the electroporation chamber distributes the current equally among the cells and maintains a stable pH throughout the chamber; these key benefits increase cell viability dramatically.
Gibco StemScale PSC Suspension Medium is a scalable, easy-to-use medium that supports large-scale generation of human pluripotent stem cells (hPSCs) in suspension culture, and formulated to address the current technical challenges in aggregating hPSC spheroid culture systems.
As you move your genome editing research from basic research to clinical settings, high-quality cell therapy ancillary materials and documentation are critical. We now offer our Gibco CTS TrueCut Cas9 Protein for clinical research that requires next generation Cas9 nuclease that is manufactured with stringent specifications.
The Invitrogen TrueCut HiFi Cas9 Protein is a high fidelity Cas9 variant engineered to improve specificity by significantly reducing off-target effects while maintaining high on-target editing efficiency.
Our high-quality products for lipid-based transfection and electroporation are useful across a wide variety of cell types and applications. Invitrogen Lipofectamine reagents are the most-cited transfection reagents in the world, with more than 300,000 citations and over 30 years of performance leadership.
Download our full product brochure or learn more about our transfection solutions here
Download the free 66 pages resource guide for key products and service to support your stem cell research.
Review our complete 63 pages research services handbook to see our range of stem cell and other cell biology services
Thanks to Microsoft HoloLens 2, Thermo Fisher Scientific is blending the virtual and physical with mixed reality to create an immersive experience to training and post-sales support. See challenges with fresh eyes, upskill your team, troubleshoot, and optimize research faster and smarter.
The Thermo Fisher Scientific Aspire member program* is a rewards program to show our appreciation, and support your contributions to science.
Join our rewards program today and start receiving points for products you use or purchase. All members can enjoy the following benefits:
* Terms, conditions, and restrictions apply to all benefits. Click to view.
** Eligible participants will receive points when enrolling. Points will be added to the Aspire program member's dashboard once the participant's qualification has been verified. All terms and conditions of the Aspire member program apply.
The Gibco CTS Rotea Counterflow Centrifugation System is a closed, cell-processing system for cell therapy that offers exceptional cell recovery, flexible input and output volume capability, and high-throughput rate. The compact footprint, process flexibility, and sterile single-use kits allow the system to seamlessly scale with your process, from research, through clinical manufacturing.
The Gibco CTS Xenon Electroporation System is a closed, scalable, electroporation system for GMP-compliant cell therapy manufacturing. It offers reliable delivery of DNA, RNA, proteins, and other molecules into cells, with exceptional cell viability and recovery, leading to efficient transfection and genome editing, even in hard-to-transfect cells. Offering high performance in volumes of 1 mL or 5–25 mL, the CTS Xenon system can be used to seamlessly scale your cell therapy workflow from process development through clinical manufacturing.
The Neon NxT Electroporation System is an electroporation machine that uses an electronic pipette tip as an electroporation chamber. The design and performance of the electronic pipette transfection chamber result in increased cell viability and transfection efficiency compared to traditional cuvette-based electroporation systems.
The proprietary tip technology of Neon NxT Electroporation System enables scientists to reduce sample transfer loss while simplifying their workflow. The system was designed for electroporation of mammalian cells, but some customers have found it to be successful for other cell types such as insect cell cultures and parasites.
The advantages of this electroporation device are:
The design of the electroporation chamber distributes the current equally among the cells and maintains a stable pH throughout the chamber; these key benefits increase cell viability dramatically.
Gibco StemScale PSC Suspension Medium is a scalable, easy-to-use medium that supports large-scale generation of human pluripotent stem cells (hPSCs) in suspension culture, and formulated to address the current technical challenges in aggregating hPSC spheroid culture systems.
As you move your genome editing research from basic research to clinical settings, high-quality cell therapy ancillary materials and documentation are critical. We now offer our Gibco CTS TrueCut Cas9 Protein for clinical research that requires next generation Cas9 nuclease that is manufactured with stringent specifications.
The Invitrogen TrueCut HiFi Cas9 Protein is a high fidelity Cas9 variant engineered to improve specificity by significantly reducing off-target effects while maintaining high on-target editing efficiency.
Our high-quality products for lipid-based transfection and electroporation are useful across a wide variety of cell types and applications. Invitrogen Lipofectamine reagents are the most-cited transfection reagents in the world, with more than 300,000 citations and over 30 years of performance leadership.
Download our full product brochure or learn more about our transfection solutions here
Download the free 66 pages resource guide for key products and service to support your stem cell research.
Review our complete 63 pages research services handbook to see our range of stem cell and other cell biology services
Thanks to Microsoft HoloLens 2, Thermo Fisher Scientific is blending the virtual and physical with mixed reality to create an immersive experience to training and post-sales support. See challenges with fresh eyes, upskill your team, troubleshoot, and optimize research faster and smarter.
The Thermo Fisher Scientific Aspire member program* is a rewards program to show our appreciation, and support your contributions to science.
Join our rewards program today and start receiving points for products you use or purchase. All members can enjoy the following benefits:
* Terms, conditions, and restrictions apply to all benefits. Click to view.
** Eligible participants will receive points when enrolling. Points will be added to the Aspire program member's dashboard once the participant's qualification has been verified. All terms and conditions of the Aspire member program apply.
For Research Use Only. Not for use in diagnostic procedures.