Search
To culture peripheral blood lymphocytes choose PB-MAX™ culture media from GIBCO® for best in class quality and performance.
PB-MAX™ karyotyping media is a fully supplemented, RPMI-based medium optimized for the karyotype analysis of peripheral blood lymphocytes. PB-MAX™ is provided as a 1X frozen liquid medium which is complete and ready to use once thawed. With no need to supplement you reduce end-user handling, thereby saving time and ensuring product integrity.
Proven for performance and reliability, each lot of PB-MAX is tested on primary peripheral blood lymphocytes and evaluated for mitotic index and chromosome banding resolution.
For Research Use Only. Not for use in diagnostic procedures.