Search
Checkweighers can provide many benefits to your process and help to make your business more efficient, reliable and profitable. The comparison tables below can help you determine which system is best for your business based on commonly defined criteria. For assistance in determining the best fit for your needs or to speak with a specialist, click the Contact Sales button above.
Package Weight Range |
|||
10g–3kg; 6kg |
35g–11kg |
10g–3kg or 10kg |
2g–300g (slack); 10g–500g (PWT) |
Max Product Rate |
|||
250–500 per min |
500–700+ per min |
250–500 per min |
250-700 per min |
Accuracy (Standard Deviation per R51/Handbook 44 Equiv.) Actual results dependent on line speed, package material/fill/weight, and handling |
|||
Up to 0.125g |
Up to 0.5g |
Up to 0.125g |
Up to 50mg |
Max Line Speed |
|||
60m/196 ft to 120m/393 ft per min |
120m/393 ft + per min |
60m/196 ft to 120m/393 ft per min |
60m/196 ft to 120m/393 ft + per min |
Package Types Supported |
|||
Rigid (cans, cardboard packages, plastic, glass) |
|||
|
|
||
Soft (plastic, paper, foil) |
|||
|
|
|
|
Configurable (multiple in or outfeeds, transfers, remote controller mounting) |
|||
|
|
|
|
Conveyor Transport Type: Slackbelt, PWT or Chain |
|||
PWT |
Chain, Chain+Belt |
PWT |
Slack |
Communications and Stats Capability |
|||
|
|
|
|
Configurable (various infeeds, outfeeds, accelerating stages, etc.) |
|||
|
|
|
|
Combos with Metal Detector or X-ray System? |
|||
|
|
|
|
Wet and/or Dry Applications |
|||
D |
W+D |
W+D |
D |
| Multi-lane Available | |||
|
|
|
|
|
||
Package Weight Range |
||
10g-3kg; 6kg
|
2g-5kg; 10g-5kg |
10g-3kg or 10kg |
Max Product Rate |
||
250–500 per min |
250–500 per min |
250–500 per min |
Accuracy (Standard Deviation per R51/Handbook 44 Equiv.) Actual results dependent on line speed, package material/fill/weight, and handling |
||
Up to 0.125g |
Up to 50mg |
Up to 0.125g |
Max Line Speed |
||
60m/196 ft–120m/393 ft per min |
60m/196 ft–120m/393 ft per min |
60m/196 ft–120m/393 ft per min |
Package Types Supported |
||
Rigid (cans, cardboard packages, plastic, glass) |
||
|
|
|
Soft (plastic, paper, foil) |
||
|
|
|
Configurable (multiple in or outfeeds, transfers, remote controller mounting) |
||
|
|
|
Conveyor Transport Type: Slackbelt, PWT or Chain |
||
PWT |
Slackbelt, or PWT |
PWT |
Communications and Stats Capability |
||
|
|
|
Configurable (various infeeds, outfeeds, accelerating stages, etc.) |
||
|
|
|
Combos with Metal Detector or X-ray System? |
||
|
|
|
Wet and/or Dry Applications |
||
D |
D |
W+D |
Configurable |
||
|
|
|
CFR 21 Part 11 Compliant |
||
|
|
|
Pharma Validation Package Available |
||
|
|
|
Package Weight Range |
||
35g-11kg |
40g-1.5kg |
2g-300g; 10g-500g |
Max Product Rate |
||
500-700+ per min |
500-700+ per min |
250-500 per min |
Accuracy (Standard Deviation per R51/Handbook 44 Equiv.) |
||
To 0.5g |
To 0.25g |
To 50mg |
Max Line Speed |
||
120m/393 ft+ per/min |
120m/393 ft+ per/min |
60m/196 ft to 120m/ 393 ft per min |
Package Types Supported |
||
Rigid (cans, cardboard packages, plastic, glass) |
||
|
Cans or plastic trays only |
|
Soft (plastic, paper, foil) |
||
|
|
|
Configurable (multiple in or outfeeds, transfers, remote controller mounting) |
||
|
|
|
Conveyor Transport Type: Slackbelt, PWT or Chain |
||
Chain, Chain+Belt |
SKATE |
Slack; PWT |
Communications and Stats Capability |
||
|
|
|
Configurable Multiple Lanes |
||
|
|
|
Combos with Metal Detector or X-ray system? |
||
|
|
|
Wet and/or Dry Applications |
||
W+D |
W+D |
D |