Invitrogen Alexa Fluor 594 dye is a bright, red-fluorescent dye that can be excited using the 561 nm or 594 nm laser lines. For stable signal generation in imaging and flow cytometry, Alexa Fluor 594 dye is pH-insensitive over a wide molar range.
Probes with high fluorescence quantum yield and high photostability allow detection of low-abundance biological structures with great sensitivity. Alexa Fluor 594 dye molecules can be attached to proteins at high molar ratios without significant self-quenching, enabling brighter conjugates and more sensitive detection.
We offer Alexa Fluor 594 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection. In addition, reactive dye forms and protein labeling kits are available to allow you to generate your own antibody conjugates or probes.
Alexa Fluor 594 dye dashboard
Initial brightness |
Direct replacement for Invitrogen Texas Red dye with bright signal and excellent photostability, especially in fixed cells; commonly used with Invitrogen Alexa Fluor 350, 488, and 647 dyes for multiplexing. In addition, is used in a range of SRM applications including dSTORM, SIM, and STED and in two-photon excitation (TPE) microscopy. | ||
Photostability in buffer |
|||
Photostability in antifade |
|||
Performance in SRM |
| 561/594 | Texas Red | 590 | 617 | ||
Laser line |
Common filter set |
Excitation max |
Alexa Fluor 594 products
We offer a broad range of Alexa Fluor 594 products including labeling kits, secondary antibodies, bioconjugates, and more:
- Super-resolution microscopy (SRM) applications
- Antibody labeling kits for direct detection
- Primary antibodies for direct detection
- Secondary antibodies for flexible labeling
- Streptavidin conjugates for signal amplification
- Tyramide labeling kits for ultimate sensitivity
- Phalloidin conjugates for actin staining
- Annexin V conjugates for apoptosis imaging
- Hydrazides for neuronal tracing
- Labeled nucleotides for hybridization probes
Microtubules of fixed bovine pulmonary artery endothelial cells were visualized with mouse monoclonal anti–α-tubulin antibody, followed by an Alexa Fluor 350 goat anti–mouse IgG antibody. Next, the F-actin was labeled with Alexa Fluor 594 phalloidin. Finally, the cells were incubated with Alexa Fluor 488 wheat germ agglutinin to stain components of endosomal pathways. The superimposed and pseudocolored images were acquired sequentially using bandpass filter sets appropriate for DAPI, Texas Red dye, and fluorescein, respectively.
Alexa Fluor 594 reactive dyes
Select dye-labeling chemistries for conjugation with the following reactive groups:
Alexa Fluor 594 protein/antibody labeling kits
Select an optimized labeling kit for your target antibody or protein:
| Target protein | Kit | Cat. No. |
|---|---|---|
| Site-specific antibody labeling | ||
| <1–20 µg mouse IgG1 | Zenon Alexa Fluor 594 Mouse IgG1 Labeling Kit | Z25007 |
| <1–20 µg mouse IgG2a | Zenon Alexa Fluor 594 Mouse IgG2a Labeling Kit | Z25107 |
| <1–20 µg mouse IgG2b | Zenon Alexa Fluor 594 Mouse IgG2b Labeling Kit | Z25207 |
| <1–20 µg human IgG | Zenon Alexa Fluor 594 Human IgG Labeling Kit | Z25407 |
| <1–20 µg rabbit IgG | Zenon Alexa Fluor 594 Rabbit IgG Labeling Kit | Z25307 |
| Conventional antibody/protein labeling | ||
| 10–20 µg antibody | APEX Alexa Fluor 594 Antibody Labeling Kit | A10474 |
| 20–100 µg antibody/protein | Alexa Fluor 594 Microscale Antibody/Protein Labeling Kit | A30008 |
| 100 µg antibody/protein | Alexa Fluor 594 Antibody/Protein Labeling Kit | A20185 |
| 1 mg antibody/protein | Alexa Fluor 594 Antibody/Protein Labeling Kit | A10239 |
Alexa Fluor dye family
Illuminating research for over 15 years
Alexa Fluor dyes are among the most trusted fluorescent dyes available today. In addition to superior performance, you’ll get a technical support team experienced in problem-solving, >50,000 published references, application and experimental tips, and protocols to help with experimental planning. For an outstanding research experience, choose Alexa Fluor dyes.
- Learn more about the brightness of Alexa Fluor dyes
- Learn more about the photostability of Alexa Fluor dyes
- Read all about Alexa Fluor dyes in the Molecular Probes Handbook
For Research Use Only. Not for use in diagnostic procedures.