MEM Vitamin Solution (100X)
Product Image
Gibco™
MEM Vitamin Solution (100X)
Gibco™ MEM Vitamin Solution is used as a growth supplement for cell culture medium, to increase cell growth and viability.Read more
Have Questions?
Catalog NumberQuantity
11120052100 mL
Catalog number 11120052
Price (USD)
23.01
Special offer
Online exclusive
Ends: 27-Dec-2025
30.68
Save 7.67 (25%)
Each
Add to cart
Quantity:
100 mL
Price (USD)
23.01
Special offer
Online exclusive
Ends: 27-Dec-2025
30.68
Save 7.67 (25%)
Each
Add to cart
Gibco™ MEM Vitamin Solution is used as a growth supplement for cell culture medium, to increase cell growth and viability. Gibco™ MEM Vitamin Solution is formulated to contain 100x the vitamins found in the standard Minimum Essential Medium (MEM). The complete formulation is available.
Dual-site cGMP Manufacturing and Quality System
Gibco™ MEM Vitamin Solution is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical Gibco™ MEM Vitamin Solution product made in our Scotland facility (11120-037). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell TypeMammalian Cells
Concentration100 X
Quantity100 mL
Shelf Life12 Months
Shipping ConditionDry Ice
FormLiquid
Product TypeVitamin Supplement
SterilitySterile-filtered
pH5.4 to 7.7
Unit SizeEach
Contents & Storage
Storage conditions: -5 to -20°C
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture
Frequently asked questions (FAQs)
What is the best way to handle the MEM Vitamin Solution (100X)? It comes frozen, so should I thaw it, then aliquot it and freeze the aliquots for later use?
If you're going to be using small batches of the vitamin solution at a time, it would be best to aliquot it out and store at -20 degrees C. The stability of the MEM Vitamin Solution is one year at -20 degrees C, while it is only 2 weeks at 4 degrees C.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the solvent used for MEM Vitamin Solution (100X)?
The solvent is distilled water.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is Gibco MEM Vitamin Solution?
The MEM Vitamin Solution (Cat. No. 11120-052 or 11120-037 depending on your country) is a growth supplement for cell culture medium. This solution is formulated to contain 100X the vitamins found in standard MEM. Find the complete formulation here (http://www.thermofisher.com/us/en/home/technical-resources/media-formulation.163.html).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.