Cell Freezing Media
Protect your frozen cells with Gibco cryopreservation media
Freezing down cells guards against unplanned mutations, senescence, and contamination. While unprotected cell freezing is ordinarily lethal, cell freezing with cryopreservation media helps protect cells for long-term viability as well as minimizes freezing injury for an intact cell supply on your schedule.
View protocols for cell freezing and thawing for more information.
Freezing down cells in DMSO 
The use of a freezing medium should contain a cryoprotective agent like dimethyl sulfoxide (DMSO) or glycerol. These agents reduce culture-medium freezing points and allow for slower cooling rates; greatly reducing the risk of ice crystal formation which can damage and kill untreated cells.
While each of the following Gibco freezing media are optimized for different cell types, they are all ready-to-use and contain 10% DMSO.
Choose the right cryopreservation media for your cell type
 Synth-a-Freeze Cryopreservation MediumRecovery Cell Culture Freezing MediumPSC Cryopreservation Kit
Tested cell typesHuman keratinocytes, ESCs, MSCs, NSCs, other primary cell typesCHO-S, CHO-K1, HEK 293, Jurkat, NIH 3T3Pluripotent stem cells, peripheral blood mononuclear cells
Not suitable forMelanocytes
Chemical propertiesAnimal origin-freeContains FBSXeno-free (Cryomedium)
Animal origin-free (RevitaCell)
Recovery Cell Culture Freezing Medium product bottle
Recovery Cell Culture Freezing Medium
A complete, ready-to-use freezing medium, the Recovery medium formulation avoids messy mixing. Using an optimized version of the classic DMEM, serum, and DMSO, Recovery cell freezing medium is ready to go when you are. When treated with Recovery medium, cells experience improved recovery and viability post-thaw.
Comparative analysis of Recovery Cell Culture Freezing Medium vs. currently available cryopreservation technologies
Bar chart of cell recovery percent for 4 different cell lines frozen in Recovery medium and 6 alternative media
Method: For each of four different cell lines (i.e., CHO, MDCK, Swiss 3T3, HeLa), the same number of cells were frozen into each of seven different cryopreservation media. Upon thawing, each cell sample was diluted into the same volume of a growth medium and seeded in 12-well plates. The initial seeding densities were 2.3×104 cells/well for adherent cell lines, and 6.0×105 cells/well for non-adherent cell lines, based on cell counts determined at the cryopreservation step. 
When the leading culture for a particular cell line had reached late log growth and none of the other cultures from that cell line had passed peak density, all cultures for that cell line were harvested and the data were analyzed. Thus, different cell lines were analyzed on different days: CHO, day 6; MDCK, day 6; Swiss 3T3, day 5; and HeLa, day 6. Cell recovery of replicate cultures from different cryopreservation media were normalized to Recovery Cell Culture Freezing Medium and plotted (average normalized recovery±standard error).
Result: For cryopreservation of both adherent and suspension cell lines, Recovery freezing medium results in an average of 25% higher cell viability compared to other cell freezing media:
Flyer: Recovery freezing medium cell viability
Synth-a-Freeze medium product bottle
Synth-a-Freeze Cryopreservation Medium
Formulated without antibiotics, antimycotics, hormones, growth factors, serum or proteins, Synth-a-Freeze medium is defined and ready-to-use. Synth-a-Freeze medium is buffered with HEPES and sodium bicarbonate and designed for use with primary mammalian cell types including human keratinocytes, embryonic stem cells, neural stem cells, and mesenchymal stem cells.
NOTE: Synth-a-Freeze medium is not appropriate for melanocytes.
PSC Cryopreservation Kit product bottles
PSC Cryopreservation Kit
The PSC Cryopreservation Kit contains ready-to-use, xeno-free cryopreservation medium as well as a bottle of RevitaCell Supplement (100X). Together, these reagents minimize cell viability loss and unwanted cell differentiation while maximizing post-thaw recovery. Applications include pluripotent stem cells cultured in media systems, such as Essential 8 and StemPro hESC SFM, as well as other commercially available formulations. In addition, this kit can also be used to cryopreserve and recover peripheral blood mononuclear cells (PBMCs).
RevitaCell supplement product bottle
RevitaCell supplement
Containing Rho-associated protein kinase (ROCK) inhibitors and antioxidant properties, the RevitaCell supplement typically recovers pluripotent stem cells at 40,000 viable cells/cm2, and significantly reduces the average post-thaw recovery time of most cryopreservation solutions. In addition to pluripotent stem cells, RevitaCell supplements can also be utilized with primary cells including neurons, human epidermal keratinocytes, and human corneal epithelial cells.
cell culture media icon GMP manufacturing
Gibco cell culture products are manufactured in facilities compliant with current good manufacturing practices (GMP) and adhere to a robust quality management system, helping to ensure the consistency, reliability, and high quality you can rely on.
Learn more about GMP manufacturing
leaf icon Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Learn more about sustainable solutions
Resources
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Cell Culture Basics
Learn best practices and fundamentals of cell culture. Topics include aseptic technique and methods for the growth and maintenance of cells in culture.
Cell Culture Videos
Explore this collection of cell culture videos providing valuable insights into techniques, resources, and stay up to date with the latest advancements in the field.
Cell Culture Selection Tools
Find the right cell culture products for your research with these selection tools. Search for media, FBS, plastics, transfection reagents, and more.
Support
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? 
Contact our expert team for technical and application support of Laboratory Products
Related Products
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments