Search
QuantiGene Plex assays provide a fast and high-throughput solution for targeted multiplexed gene expression quantitation, allowing for the simultaneous measurement of up to 80 genes of interest in a single well of a 96- or 384-well plate. This “lyse-and go” solution does not require RNA purification and works for a variety of sample types including cultured cells, fresh frozen tissue, formalin-fixed paraffin-embedded (FFPE) tissue specimens, and whole blood samples.
Review our Biomarker quantitation assay guide
QuantiGene Gene Expression guides and tools | Featured applications using Invitrogen QuantiGene Gene Expression Profiling products |
QuantiGene Plex Assays incorporate branched DNA technology and Luminex xMAP technology for accurate multiplex gene expression profiling. Branched DNA assays allow for the direct measurement of RNA transcripts by using signal amplification rather than target amplification.
Invitrogen QuantiGene SinglePlex assays provide a fast and high-throughput solution for gene expression quantitation, allowing the measurement of a single gene of interest per well of a 96- or 384-well plate
Let Thermo Fisher Scientific help you in designing your preferred and customized QuantiGene Plex panel, run your samples, and deliver your data within 30 days. We accept: cell lysates, whole blood lysates, tissue homogenates, and purified RNA.
Analyze both protein and gene expression from a single sample. Invitrogen ProcartaPlex and QuantiGene Plex multiplex quantitation assays can all be used on any Luminex Instrument.
Discover how multiplexing can be used to examine secreted proteins and mRNA expression levels on a single Luminex platform using a single sample. This Methods paper specifically details how the ProcartaPlex and QuantiGene Plex assays can be paired to measure both proteins and messenger RNA (mRNA) expression in a single sample run on a Luminex platform