Search
For applications requiring stirring capacities of 20L to 600L, we offer multiple models for both general lab operating conditions and special operating conditions.
Find the large capacity hot plate stirrer to meet your needs using our hot plate stirrer selection guide ›
| Cimarec Biosystem Slow-Speed Stirrer for Cell Culture | |
|---|---|
![]() | |
| Stir thermally sensitive cultures including microcarrier cultures, culture broths and cell suspensions with the Thermo Scientific Cimarec Biosystem Slow-Speed Stirrer for Cell Culture. This product offers gentle stirring and low heat output to protect sensitive cell cultures within CO2 incubators. | |
| Part no. | 50119113 |
| Plate material | Stainless steel |
| Plate shape | Square |
| Plate size | 7 x 7 in. (18 x 18cm) |
| Watts | 4w |
| Speed | 5 to 120rpm |
| Capacity | 20L |
| Operating Conditions | -10° to +56°C at 100% humidity, 0° to +50°C in a waterbath |
| Cimarec Mobil 10 and Mobil 25 Large Volume Stirrer | ||
|---|---|---|
![]() | ||
| Stir and mix volumes of up to 10L with Thermo Scientific Cimarec Mobil 10 and Mobil 25 Large Volume Stirrers, ideal for hard-to-stir viscous liquids. The “Quick-Stop” function stops the drive within three seconds, minimizing after runs and reducing the risk of breakage. | ||
| Part no. | 50119111 | 50119112 |
| Model | Mobil 10 w/o controller | Mobil 25 w/o controller |
| Plate material | Stainless steel | Stainless steel |
| Plate shape | Round | Round |
| Plate size | 5 in. (128mm) dia. | 5 in. (128mm) dia. |
| Voltage | NA | NA |
| Speed | 100 to 1000rpm | 100 to 1000rpm |
| Capacity | 20L | 25L |
| Operating Conditions | -10° to 56°C | -10° to 121°C |
| Cimarec Power Direct High Capacity Stirrer | |
|---|---|
![]() | |
| Stir up to 40L with the integrated controller of the Thermo Scientific Cimarec Power Direct, ideal for high-capacity stirring. | |
| Part no. | 50098760 |
| Plate material | Stainless steel |
| Plate shape | Rectangular |
| Plate size | 11.6 x 9.4 in. (295 x 240mm) |
| Voltage | 100-240V |
| Speed | 100 to 2000rpm |
| Capacity | 40L |
| Operating Conditions | -10° to 40°C |
| Cimarec Mobil Direct Stirrers | ||||||
|---|---|---|---|---|---|---|
![]() | ||||||
| Perform high-volume applications with the exceptional stirring power of Thermo Scientific Cimarec Mobil Direct Stirrers. The Mobil 200 and 600 require either the Telemodul 40 M or the Telemodul 80 M control unit. | ||||||
| Part no. | 50088128 | 50088129 | 50088131 | 50100547 | 50119109 | 50119110 |
| Model | Mobil Direct | Mobil Direct | Mobil Direct | Mobil Direct | Mobil 200 without Controller | Mobil 600 without Controller |
| Plate material | Stainless steel | Stainless steel | Stainless steel | Stainless steel | Stainless steel | Stainless steel |
| Plate shape | Square | Square | Square | Square | Square | Square |
| Plate size | 13 x 13 in. (330 x 330mm) | 13 x 13 in. (330 x 330mm) | 13 x 13 in. (330 x 330mm) | 13 x 13 in. (330 x 330mm) | 7 x 7 in. (80 x 80mm) | 13 x 13 in. (330 x 330mm) |
| Voltage | 230V | 100V | 115V | 230V | NA | NA |
| Plug Type | Euro | Japan | USA | UK | NA | NA |
| Speed | 100 to 1000rpm | 100 to 1000rpm | 100 to 1000rpm | 100 to 1000rpm | 100 to 1000rpm | 100 to 1000rpm |
| Capacity | 150L | 150L | 150L | 150L | 200L | 600L |
| Operating Conditions | -10° to +40°C at 95% rH | -10° to +40°C at 95% rH | -10° to +40°C at 95% rH | -10° to +40°C at 95% rH | -10° to +56°C at 100% rH | -10° to +56°C at 100% rH |
| RT2 Basic Hotplate Stirrer | ||
|---|---|---|
![]() | ||
| Rely on safety and consistent stirring speed with the Thermo Scientific RT2 Basic hotplate stirrer. Benefit from exceptionally strong magnet coupling power and a smooth-start stirring mechanism. Experience fast heat-up time and an excellent heat transfer rate thanks to the tightly integrated structure of the heater and a ceramic-coated aluminum alloy top plate which provides excellent temperature uniformity and corrosion resistance while allowing for optimal monitoring of color changes in samples or media during operation. | ||
| Part no. | 88880003 | 88880004 |
| Plate material | Ceramic-coated aluminum | Ceramic-coated aluminum |
| Plate shape | Round | Round |
| Plate size | 5.5 in. (140mm) dia. | 5.5 in. (140mm) dia. |
| Voltage | 230V | 120V |
| Speed | 50 to 2,000rpm | 50 to 2,000rpm |
| Temperature Range | 50° to 350°C | 50° to 350°C |
| Capacity | 20L | 20L |
| 5.5 in. (140mm) dia. |
| RT2 Advanced Hotplate Stirrer | ||
|---|---|---|
![]() | ||
| It’s all about precise control without sacrificing your budget with the sleek, low profile Thermo Scientific RT2 Advanced Digital Hotplate Stirrer which offers digital temperature and speed control with 0.1°C temperature resolution and stir speed range of 30 to 2000rpm. Choose your heat-up rate and experience an excellent heat transfer due to the tightly integrated structure of the heater and a ceramic-coated aluminum alloy top plate that also provides excellent uniformity and corrosion resistance while allowing for optimal monitoring of color changes in samples or media during operation. | ||
| Part no. | 88880006 | 88880005 |
| Plate material | Ceramic-coated aluminum | Ceramic-coated aluminum |
| Plate shape | Round | Round |
| Plate size | 5.5 in. (140mm) dia. | 5.5 in. (140mm) dia. |
| Voltage | 120V | 230V |
| Speed | 30 to 2,000rpm | 30 to 2,000rpm |
| Temperature Range | 50° to 350°C | 50° to 350°C |
| Capacity | 20L | 20L |
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.