Search
Our multi-point magnetic stirrers are available with 4, 6, 15 or 60 stirring points with good speed control to meet various needs in all labs. They are applicable in different operating conditions such as daily lab use, water baths and CO2 incubators.
Find the stirrer to meet your needs using our stirrer selection guide ›
| Super-Nuova Multi-Position Digital Stirrers | ||
|---|---|---|
![]() | ||
| Carry out solubility studies, digestion, titration and enzymatic studies with Thermo Scientific Super-Nuova Multi-Position Digital Stirrers. Offering four individually-controlled stirring positions, ceramic tops and 4-function memory, these stirrers are designed for stronger coupling and stirring control with advanced safety features. Ideal for labs with multiple users. | ||
| Part no. | S136035Q | S136030-33Q |
| Plate material | Ceramic | Ceramic |
| Plate shape | Square | Square |
| Plate size | 10.25 x 10.25 in. (26.0 x 26.0cm) | 10.25 x 10.25 in. (26.0 x 26.0cm) |
| Voltage | 120V | 220/240V |
| Speed | 50 to 1,200rpm | 50 to 1,200rpm |
| Stirring positions | 4 | 4 |
| Cimarec i Poly 15 and Multipoint Stirrers | |||
|---|---|---|---|
![]() | |||
| Conduct stirring applications with Thermo Scientific™ Cimarec™ i Poly 15 and Multipoint Stirrers for reliable, safe performance. These stirrers are 100% maintenance- and wear-free, in a fully synchronized multipoint platform. | |||
| Part no. | 50093557 | 50093538 | 50094596 |
| Model | Multipoint 6 | Multipoint 15 | Poly 15 |
| Plate material | Stainless steel | Stainless steel | Stainless steel |
| Plate shape | Rectangular | Rectangular | Rectangular |
| Plate size | 16.5 x 9.4 in. (420 x 240mm) | 16.5 x 9.4 in. (420 x 240mm) | 16.5 x 9.4 in. (420 x 240mm) |
| Voltage | 100-240V | 100-240V | 100-240V |
| Speed | 80 to 2000rpm | 80 to 2000rpm | 130 to 990rpm |
| Stirring positions | 6 | 15 | 15 |
| Operating Conditions | -5° to +40°C at 95% rH | -5° to +40°C at 95% rH | -5° to +40°C at 95% rH |
| Cimarec Biosystem Slow-Speed Stirrer for Cell Culture | |
|---|---|
![]() | |
| Stir thermally sensitive cultures including microcarrier cultures, culture broths and cell suspensions with the Thermo Scientific Cimarec Biosystem Slow-Speed Stirrer for Cell Culture. This product offers gentle stirring and low heat output to protect sensitive cell cultures within CO2 incubators. | |
| Part no. | 50119114 |
| Plate material | Stainless steel |
| Plate shape | Square |
| Plate size | 12.9 x 12.9 in. (33 x 33cm) |
| Watts | 4w |
| Speed | 5 to 120rpm |
| Stirring positions | 4 |
| Operating Conditions | -10 to +56°C at 100% rH |
| Cimarec i Telesystem Multipoint Stirrers | ||||||
|---|---|---|---|---|---|---|
![]() | ||||||
| Safely conduct applications with aggressive media in high-humidity environments with Thermo Scientific Cimarec i Telesystem Multipoint Stirrers. The stirrers are designed for integration with incubators and controlled climate chambers. | ||||||
| Part no. | 50088077 | 50088078 | 50088034 | 50088036 | 50088009 | 50088011 |
| Model | Telesystem 6 Position w/Telemodul 20 C | Telesystem 6 Position w/Telemodul 40 C | Telesystem 15 Position w/Telemodul 20 C | Telesystem 15 Position w/Telemodul 40 C | Telesystem 60 Position w/Telemodul 20 C | Telesystem 60 Position w/Telemodul 40 C |
| Plate material | Stainless steel | Stainless steel | Stainless steel | Stainless steel | Stainless steel | Stainless steel |
| Plate shape | Rectangular | Rectangular | Rectangular | Rectangular | Rectangular | Rectangular |
| Plate size | 16.5 x 9.4 in. (420 x 240mm) | 16.5 x 9.4 in. (420 x 240mm) | 16.5 x 9.4 in. (420 x 240mm) | 16.5 x 9.4 in. (420 x 240mm) | 16.5 x 9.4 in. (420 x 240mm) | 16.5 x 9.4 in. (420 x 240mm) |
| Voltage | 100-240V | 100-240V | 100-240V | 100-240V | 100-240V | 100-240V |
| Speed | 130 to 1400rpm | 100 to 2000rpm | 130 to 1400rpm | 100 to 2000rpm | 130 to 1400rpm | 100 to 2000rpm |
| Stirring positions | 6 | 6 | 15 | 15 | 60 | 60 |
| Operating Conditions | -10° to +56°C at 100%RH in air; 0° to 50°C submerged in water | |||||
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.