Search
Single point magnetic stirrers are designed with various speed ranges for use in all labs. Basic, standard and advanced stirrers are designed for general lab operating conditions, and specialty stirrers are used for specific operating conditions such as submersion in water or in CO2 incubators.
Find the stirrer to meet your needs using our stirrer selection guide ›
| Cimerac Basic Stirrers | ||
|---|---|---|
![]() | ||
| Reach stirring speeds from 250 to 2500rpm with Thermo Scientific Cimarec Basic Stirrers, ideal for economical, general-purpose use. | ||
| Part no. | S194615 | S194925 |
| Plate material | Ceramic | Ceramic |
| Plate shape | Square | Square |
| Plate size | 4.25 x 4.25 in. (10.8 x 10.8cm) | 7.25 x 7.25 in. (18.4 x 18.4cm) |
| Voltage | 120V | 120V |
| Speed | 250 to 2500rpm | 250 to 2500rpm |
| RT Basic Series Magnetic Stirrers | ||||||
|---|---|---|---|---|---|---|
![]() | ||||||
| Experience reliability with the plug-and-play Thermo Scientific RT Basic Series Magnetic Stirrers, ideal for routine stirring applications. Three size options to choose from—120mm, 170mm, or 220mm—all with a low profile lightweight design with an extremely small footprint. | ||||||
| Part no. | 88880007 | 88880008 | 88880009 | 88880010 | 88880011 | 88880012 |
| Plate material | Polypropylene | Polypropylene | Polypropylene | Polypropylene | Polypropylene | Polypropylene |
| Plate shape | Round | Round | Round | Round | Round | Round |
| Plate size | 4.72 in. (120mm) dia. | 4.72 in. (120mm) dia. | 6.69 in. (170mm) dia. | 6.69 in. (170mm) dia. | 8.66 in. (220mm) dia. | 8.66 in. (220mm) dia. |
| Voltage | 100-240V | 100-240V | 100-240V | 100-240V | 100-240V | 100-240V |
| Plug type | USA | Cord set w/various plugs | USA | Cord set w/various plugs | USA | Cord set w/various plugs |
| Speed | 150 to 2500RPM | 150 to 2500RPM | 150 to 2500RPM | 150 to 2500RPM | 150 to 2500RPM | 150 to 2500RPM |
| Cimarec+ Stirrer Series | ||||
|---|---|---|---|---|
![]() | ||||
| Our Thermo Scientific Cimarec+ series digital stirrers offer precise stirring control for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operations. | ||||
| Part no. | ||||
| Plate material | Ceramic | Ceramic | Aluminum | Ceramic |
| Plate shape | Square | Square | Square | Square |
| Plate size | 4.25 x 4.25 in. (10.8 x 10.8cm) | 7.25 x 7.25 in. (18.4 x 18.4cm) | 7.25 x 7.25 in. (18.4 x 18.4cm) | 10.25 x 10.25 in. (26.0 x 26.0cm) |
| Speed | 50 to 1500RPM | 50 to 1500RPM | 50 to 1500RPM | 50 to 1500RPM |
| RT Touch Series Magnetic Stirrers | ||||
|---|---|---|---|---|
![]() | ||||
| Stay in control with Thermo Scientific RT Touch Series Magnetic Stirrers, low profile digital display stirrers ideal for routine applications where visible stir speed is required. | ||||
| Part no. | 88880013 | 88880014 | 88880015 | 88880016 |
| Plate material | Polypropylene | Polypropylene | Polypropylene | Polypropylene |
| Plate shape | Round | Round | Round | Round |
| Plate size | 6.69 in. (170mm) dia. | 6.69 in. (170mm) dia. | 8.69 in. (220mm) dia. | 8.69 in. (220mm) dia. |
| Voltage | 100-240V | 100-240V | 100-240V | 100-240V |
| Plug Type | USA | Cord set w/various plugs | USA | Cord set w/various plugs |
| Speed | 30 to 2000rpm | 30 to 2000rpm | 30 to 2000rpm | 30 to 2000rpm |
| SuperNuova+ Stirrer Series | ||||
|---|---|---|---|---|
![]() | ||||
| Our Thermo Scientific SuperNuova+ stirrer series provides reliable performance, dependable safety and simple operation. Units are available in two sizes, including a multi-position model with optimized controls and settings for applications that demand advanced precision. | ||||
| Part no. | S88857190 | S88857195 | S88850190 | S88850195 |
| Plate material | Ceramic | Ceramic | Ceramic | Ceramic |
| Plate shape | Square | Square | Square | Square |
| Plate size | 7.25 x 7.25 in. (18.4 x 18.4cm) | 7.25 x 7.25 in. (18.4 x 18.4cm) | 10.25 x 10.25 in. (26.0 x 26.0cm) | 10.25 x 10.25 in. (26.0 x 26.0cm) |
| Voltage | 100-120V | 230V | 100-120V | 230V |
| Speed | 50 to 1500rpm | 50 to 1500rpm | 50 to 1500rpm | 50 to 1500rpm |
| Cimarec Biosystem Slow-Speed Stirrer for Cell Culture | |
|---|---|
![]() | |
| Stir thermally sensitive cultures including microcarrier cultures, culture broths and cell suspensions with the Thermo Scientific Cimarec Biosystem Slow-Speed Stirrer for Cell Culture. This product offers gentle stirring and low heat output to protect sensitive cell cultures within CO2 incubators. | |
| Part no. | 50119113 |
| Plate material | Stainless steel |
| Plate shape | Square |
| Plate size | 7 x 7 in. (18 x 18cm) |
| Watts | 4w |
| Speed | 5 to 120rpm |
| Operating conditions | -10° to +56° at 100% humidity 0° to +50° in a waterbath |
| Cimarec i Mini Stirrers | ||||||
|---|---|---|---|---|---|---|
![]() | ||||||
| Work in the smallest of places with Thermo Scientific Cimarec i Mini Stirrers. The i Mini Series stirrers come with or without your choice of controller for use with the distribution system. | ||||||
| Part no. | 50088118 | 50088120 | 50088126 | 50088130 | 50088132 | 50088140 |
| Model | Mini w/ Telemodul | Mini 20 w/ Telemodul 20 C | Mini w/ Telemodul | Mini w/ Telemodul | Mini w/ Telemodul | Mini w/o controller |
| Plate material | Stainless steel | Stainless steel | Stainless steel | Stainless steel | Stainless steel | Stainless steel |
| Plate shape | Square | Square | Square | Square | Square | Square |
| Plate size | 0.47 x 0.47 in. (12 x 12mm) | 0.47 x 0.47 in. (12 x 12mm) | 0.47 x 0.47 in. (12 x 12mm) | 0.47 x 0.47 in. (12 x 12mm) | 0.47 x 0.47 in. (12 x 12mm) | 0.47 x 0.47 in. (12 x 12mm) |
| Voltage | 115V | 100-240V | 100V | 230V | 230V | N/A |
| Plug Type | USA | Cord set w/various plugs | Japan | Euro | UK | N/A |
| Speed | 130 to 1000rpm | 130 to 1400rpm | 130 to 1000rpm | 130 to 1000rpm | 130 to 1000rpm | N/A |
| Operating conditions | -10 to +56°C at 95% rH | |||||
| Cimarec i Micro Stirrers | ||||||
|---|---|---|---|---|---|---|
![]() | ||||||
| Work with Thermo Scientific Cimarec i Micro Stirrers, hermetically sealed IP68-protection class stirrers that can be submerged in water. The Cimarec i Micro Series stirrers come standard with your choice of controller or without controller for use with distribution system. | ||||||
| Part no. | 50088148 | 50088149 | 50088150 | 50088151 | 50088155 | 50088162 |
| Plate material | Stainless steel | Stainless steel | Stainless steel | Stainless steel | Stainless steel | Stainless steel |
| Plate shape | Square | Square | Square | Square | Square | Square |
| Plate size | 1.8 x 1.8 in. (48 x 48mm) | 1.8 x 1.8 in. (48 x 48mm) | 1.8 x 1.8 in. (48 x 48mm) | 1.8 x 1.8 in. (48 x 48mm) | 1.8 x 1.8 in. (48 x 48mm) | 1.8 x 1.8 in. (48 x 48mm) |
| Voltage | 100-240V | 100V | 115V | 230V | 230V | 230V |
| Plug Type | Cord set with various plugs | Japan | USA | UK | Australia | Euro |
| Speed | 130 to 1400rpm | 130 to 1000rpm | 130 to 1000rpm | 130 to 1000rpm | 130 to 1000rpm | 130 to 1000rpm |
| Operating conditions | -10 to 100°C at 95% rH, -10 to 120°C in dry air, 0-95°C submerged in water | |||||
| Cimarec i Compact and Maxi Stirrers | |||||
|---|---|---|---|---|---|
![]() | |||||
| Handle challenging stirring environments with Thermo Scientific Cimarec i Compact and Maxi Stirrers, 100% maintenance– and wear–free hermetically sealed units. With powerful magnets and no moving parts, these inductive-drive stirrers are designed to go into water baths and incubators. The Cimarec i Compact and Maxi series stirrers come standard with your choice of controller. | |||||
| Part no. | |||||
| Model | Maxi w/ Telemodul | Maxi w/ Telemodul 20 C | Maxi w/ Telemodul 40 C | Compact w/ Telemodul | Compact w/ Telemodul 20 C |
| Plate material | Stainless steel | Stainless steel | Stainless steel | Stainless steel | Stainless steel |
| Plate shape | Square | Square | Square | Square | Square |
| Plate size | 7.9 x 7.9 in. (180 x 180mm) | 7.9 x 7.9 in. (180 x 180mm) | 7.9 x 7.9 in. (180 x 180mm) | 4.7 x 4.7 in. (120 x 120mm) | 4.7 x 4.7 in. (120 x 120mm) |
| Speed | 130 to 1000rpm | 130 to 1400rpm | 130 to 2000rpm | 130 to 1000rpm | 130 to 1400rpm |
| Operating conditions | -10 to +56°C in air; 0 to 50°C submerged in water | ||||
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.