Microarray Solutions for Companion Animals
Accelerate your unique breeding and testing programs with agrigenomics solutions for companion animals
Single nucleotide polymorphism (SNP) arrays can measure tens of thousands of SNP targets, while tandem repeat assays usually have several orders of magnitude fewer markers. This difference gives Applied Biosystems Axiom microarray results a much greater resolution and allows for verification of pedigrees and the genetic health of companion animals such as dogs, cats, and horses. The Axiom portfolio allows you to genotype entire animal populations, facilitating more tailored veterinary care and helping inform breeding efforts.
New feline genotyping array
neon-benefits-workflow
Customer and solutions overview
Understanding the feline genome with advanced microarray technology
Dr. Leslie Lyons developed a 630K SNP feline array to advance genetic research in cats, aiding both veterinary and human health studies.
Canine High-Density (HD) Genotyping Array
The Axiom Canine HD Genotyping Array is a high-throughput tool with 700,000+ SNPs and indels across 40+ breeds, ideal for canine and human disease research and breeding programs.
Advanced genetic tools for canine research
Dogs, due to their unique genetic diversity, human-like traits, and compatibility with advanced genotyping tools like Axiom Genotyping Solution, serve as powerful model organisms for both medical research and informed breeding, benefiting human and canine health alike.
Axiom Equine Genotyping Array
Equine genotyping array for whole-genome, high-density genotyping of 20 diverse worldwide breeds with over 670,796 markers.
Developments and challenges in canine genetic research
Dr. Karlsson’s Darwin’s Dogs project gathered data from 15,000 dogs using Axiom Canine Genotyping arrays to enable large-scale, cost-effective genetic research.
Agrigenomics solutions for companion animals
Choose from innovative technologies to analyze canine, equine, feline, and rabbit genomes.
For Research Use Only. Not for use in diagnostic procedures.
Custom arrays
Microarray Research Services Laboratory