Search
Western blot analysis was performed on whole cell extracts (30 µg lysate) of NTERA-2 (Lane 1), NCCIT (Lane 2), F9 (Lane 3), HEK-293 (Lane 4), A431 (Lane 5) and HeLa (Lane 6). The blots were probed with ABfinity™ Anti-OCT4 rabbit Monoclonal Antibody (Product ## 701756, 1-2 ug/ml) and detected by chemiluminescence using Goat anti-Rabbit IgG (H+L) Superclonal™ Secondary Antibody, HRP conjugate (Product ## A27036, 0.4 ug/ml, 1:2500 dilution). A 38 kDa band corresponding to OCT4 was observed only in specific cell lines. Known quantity of protein samples were electrophoresed using Novex® NuPAGE® 4-12% Bis-Tris gel (Product ## NP0321BOX), XCell SureLock™ Electrophoresis System (Product ## EI0002) and Novex® Sharp Pre-Stained Protein Standard (Product ## LC5800). Resolved proteins were then transferred onto a nitrocellulose membrane with iBlot® Dry Blotting System (Product ## IB21001). The membrane was probed with the relevant primary and secondary Antibody following blocking with 5% skimmed milk. Chemiluminescent detection was performed using Novex® ECL Chemiluminescent Substrate Reagent Kit (Product ## WP20005).