Search
Formalin-fixed paraffin-embedded (FFPE) mouse intestine was treated with DNase to fragment the DNA After treatment, the Click-iT® Plus TUNEL assay with Alexa Fluor® 594 dye (red) was used to detect the fragmented DNA. After the TUNEL reaction, actin was stained with Alexa Fluor® 647 phalloidin (purple). The cells were counterstained with Hoechst 33342 (blue). The GFP expressing epithelial cells are clearly visible (green).