Search
One Lambda™ LABType™ sequence-specific oligonucleotide (SSO) tests combine the robustness of reverse-SSO (rSSO) method typing with the Luminex platform, to provide Class I and Class II HLA typing. The assay utilizes bead technology for a streamlined workflow, able to scale the number of samples and target loci per run, delivering accurate results within hours. The multiplex analysis minimizes labor, reagent use, reaction inconsistencies, and assay time compared to a multi-well format, making it a cost-effective solution. The assay is flexible, scalable, with reliable typing and same-day results.
A suspension array platform employs fluorescently coded microspheres as a solid support to immobilize oligonucleotide probes. Target DNA is amplified with biotinylated, locus specific primers. The amplified product is then denatured and hybridized with the oligonucleotide probes. Amplified product hybridized to microsphere-bound oligonucleotide probes are labeled with R-phycoerythrin–conjugated streptavidin (SAPE) and then processed by the LABScan™ system. The assay takes place in a single well of a 96-well PCR plate.
LABType CWD assays are designed to resolve alleles as defined by the Common, Intermediate and Well Documented catalog. The kits detect HLA alleles defined for CIWD Version 3.0.0 with a broad and global scope of allele frequencies and diversity when analyzed with the Fusion CIWD filter. Loci coverage meets standards for patient HLA Typing, along with requirements for confirmatory HLA typing (HLA-CT) for unrelated donors, and enhanced alignment with LABScreen Single Antigen in determining Donor-Specific Antibody (DSA) identification.
CIWD 3.0.0 allelic resolution: Allelic coverage in alignment with the latest CIWD 3.0.0 catalog and across ethnic allele groups.
Ambiguity resolution: Improved ambiguity resolution across HLA-A, -B, -C and -DRB1 loci.
Alignment with antibody detection: The allele coverage is aligned with the specificity and compatibility of LABScreen products.
| Abbreviation | Region |
|---|---|
| AFA | African / African American |
| API | Asian/Pacific Islands |
| EURO | European / European descent |
| MENA | Middle East / North Africa |
| NAM | Native American |
| HIS | South or Central America / Hispanic / Latino |
| UNK | Unknown / Multi-ancestries |
With our LABType SSO kits, you get superior resolution and a cost-effective solution for HLA A, B, C, DRB1, DRB345, and DQA1, DQB1, DPA1, DPB1 HLA typing.
HLA Fusion™ software utilizes color-coded filtering functionality* to help you easily identify and define CIWD alleles. Additionally, you can import separate CIWD lists using a dedicated function, providing inclusion of CIWD HLAID alleles and CIWD groups for a more comprehensive analysis.
* Latest version of HLA Fusion should be downloaded for all new tools and capabilities.
LABType SSO, LABType XR/CWD, and HLA Fusion Software are CE marked in the EU and 510k-cleared in the US for in vitro diagnostic use. Availability of products in each country depends on local regulatory marketing authorization status.