Search
Join the next generation of chimerism testing with the One Lambda Devyser Chimerism NGS Assay offers a high-throughput, sensitive solution for the detection of chimerism and microchimerism.
Designed to provide accurate and sensitive detection of microchimerism levels in samples, the One Lambda Devyser Chimerism NGS Assay is an excellent solution for HLA and molecular core labs. It offers a fast and repeatable workflow from sample to report and a unified workflow. One tube per sample avoids any need for sample splitting, and the single measurement method is independent of mixed chimerism levels. Leveraging next-generation sequencing (NGS) technology, our assay offers an unparalleled limit of detection of 0.05%, enabling the identification of even the smallest changes in chimerism status.
Our One Lambda Devyser Chimerism NGS Assay offers an easy workflow that requires as little as 45 minutes of hands-on time, delivers results within 24 hours, and helps ensure maximum efficiency in your laboratory. By utilizing a single protocol, you can eliminate the need for sample-specific primers and marker tracking. The universal reagent mix allows for cost-effective testing, reducing reagent wastage and the need to keep multiple reagents in stock.
The One Lambda Devyser Chimerism NGS Assay combines the sensitivity of real-time PCR analysis and the precision of STR based analysis. The unique 24-indel panel allows for accurate detection of microchimerism, irrespective of mixed chimerism levels. With one tube per sample, there's no need for sample splitting, ensuring optimal sample integrity and reliable results.
You can easily assess chimerism, sample type, and longitudinal tracking with the One Lambda Devyser Chimerism NGS Assay and its dedicated software. The dedicated data analysis software delivers precise and speedy chimerism status information. The user-friendly interface and flexible data display simplify data interpretation. With this comprehensive sample-to-result solution, you can optimize your lab resources while providing top-quality results.
For Research Use Only. Not for use in diagnostic procedures. Availability of product in each country depends on local regulatory marketing authorization status.