PubSummary | Incidence and impact of allele-specific anti-HLA antibodies and high-resolution HLA genotyping on assessing immunologic compatibility
PubSummary | Incidence and impact of allele-specific anti-HLA antibodies and high-resolution HLA genotyping on assessing immunologic compatibility
Allele specific anti-HLA reactivity was shown to occur in around 15% of patients awaiting transplantation.
Added Date:
2021-04-11
Filters:
Filter Category Filter Values
Resource Type
  • Publication Summary
Keywords
  • single antigen beads
  • DSA
  • LABScreen Single Antigen
Products and Services
  • LABScreen Single Antigen
Scope Filters
  • Scientific Research
Areas of Interest
  • DSA
  • Single Antigen Beads
Country Availability:
()
Gated Form:
None Set