PubSummary | Discovery and Validation of a Urinary Exosome mRNA Signature for the Diagnosis of Human Kidney Transplant Rejection
PubSummary | Discovery and Validation of a Urinary Exosome mRNA Signature for the Diagnosis of Human Kidney Transplant Rejection
Analyzing urinary exosomes is a simple and effective method of post-transplant monitoring. The mRNA profiles have high predictive abilities for both any-cause rejection and T cell–mediated rejection.
Added Date:
2023-04-28
Filters:
Filter Category Filter Values
Resource Type
  • Publication Summary
Products and Services
  • LABScreen Single Antigen
Scope Filters
  • Scientific Research
Areas of Interest
  • TCMR
  • ABMR
Country Availability:
()
Gated Form:
None Set