Product Image
Invitrogen™
ELISA/ELISPOT Coating Buffer Powder
The ELISA/ELISPOT Coating Buffer Powder is designed and tested for use as the working solution for coating the ELISA orRead more
Have Questions?
Catalog NumberQuantity
00-0044-591 each
Catalog number 00-0044-59
Price (EUR)
43,89
Online exclusive
52,25
Save 8,36 (16%)
Each
Quantity:
1 each
Price (EUR)
43,89
Online exclusive
52,25
Save 8,36 (16%)
Each
The ELISA/ELISPOT Coating Buffer Powder is designed and tested for use as the working solution for coating the ELISA or ELISPOT capture antibody to the ELISA or ELISPOT plate microwells. This has been tested with all cytokine ELISA and ELISPOT capture antibodies, according to the procedures described in the corresponding protocols.
The eBioscience ELISA/ELISPOT Coating Buffer Powder is a dried powder formulation of phosphate buffered saline. Each packet is reconstituted in 1 Liter of distilled, deionized water, then filtered using a 0.22 um filter. The resulting 1x PBS (pH 7.4) is suitable for use as the ELISA/ELISPOT coating buffer.
Reported Application
ELISA, ELISPOT
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)ELISA, ELISPOT
FormPowder
Product TypeCoating Buffer
Quantity1 each
Unit SizeEach
Frequently asked questions (FAQs)
What other reagents will I need to run an ELISA using the antibody pairs?
When working with our matched pairs or uncoated ELISA kits, please refer to the user guide provided with your antibody-matched pair for kit-specific guidance. When designing your own ELISA, please utilize our ELISA Builder tool for reagent selection as well as our ELISA Development and Optimization pages for futher guidance.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.