GeneChip™ Scanner 3000 7G AutoLoader Carousel
Applied Biosystems™
GeneChip™ Scanner 3000 7G AutoLoader Carousel
The GeneChip Scanner 3000 7G AutoLoader Carousel is designed for use with the GeneChip Autoloader, which now comes pre-assembled withRead more
Have Questions?
Catalog NumberQuantity
00-00941 Ea.
Catalog number 00-0094
Price (MXN)
-
Quantity:
1 Ea.
The GeneChip Scanner 3000 7G AutoLoader Carousel is designed for use with the GeneChip Autoloader, which now comes pre-assembled with the GeneChip Scanner 3000 7G. The Autoloader allows maximum throughput and walk-away automation of the GeneChip Scanner 3000 7G in a tempature-controlled environment.
Temperature-controlled Environment
The GeneChip AutoLoader provides a temperature-controlled environment to maintain long-term sample stability and integrity for up to 16 hours. The environment for the 48-sample carousel is maintained at 15°C for all arrays, both pre-and post-scan. Extensive testing on the temperature stability of GeneChip™ arrays has shown that data concordance and assay integrity are maintained for arrays cooled at 15°C for at least 16 hours. High-resolution high-density arrays prepared with Array Holding Buffer are stable for up to 48 hours at 15°C.
48-array carousel for walk-away freedom
The AutoLoader includes a 48-array, removable carousel for unattended loading and unloading of the experiments for complete walk-away automation.
Ease of use
The AutoLoader, with its convenient, user-friendly front-loading position, is designed to maximize ease of operation with a simple user interface. This interface also shows the carousel inventory and experiments completed for a run in progress. The AutoLoader comes with an external handheld barcode reader for easy experiment creation with the information-rich Affymetrix 2D barcode.
Uninterruptible power supply (UPS)
The AutoLoader can be configured with an optional uninterruptible power supply capable of maintaining the temperature-controlled carousel environment for at least four hours in the event of a power failure.
Related products
GeneChip Scanner 3000 7G System
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)GeneChip Scanner 3000 7G with Autoloader
Product LineGeneChip
Quantity1 Ea.
Unit SizeEach